Thermo Scientific Chemicals

(S)-(+)-3-Methyl-2-Butanol, 99 %, Thermo Scientific Chemicals

Katalognummer: L09431.MC
100 mg, Each
Thermo Scientific Chemicals

(S)-(+)-3-Methyl-2-Butanol, 99 %, Thermo Scientific Chemicals

Katalognummer: L09431.MC
100 mg, Each
Menge
Katalognummer: L09431.MC
auch als L09431-MC bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
1517-66-4
IUPAC Name
3-methylbutan-2-ol
Molecular Formula
C5H12O
InChI Key
MXLMTQWGSQIYOW-UHFFFAOYNA-N
SMILES
CC(C)C(C)O
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Refractive Index
1.4080-1.4110 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid

Beschreibung

It is an pharmaceutical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Anwendungen
Es ist ein pharmazeutisches und organisches Zwischenprodukt.

Löslichkeit
Löslich in Alkohol. Unlöslich in Wasser.

Hinweise
Kühl und trocken lagern. Nicht kompatibel mit starken Oxidationsmitteln. Belüftung sicherstellen. Vor elektrostatischen Aufladungen schützen.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 3
    Acute Inhalation Toxicity - Vapors
    Category 4
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H332 - Harmful if inhaled

    H335 - May cause respiratory irritation

    Physical Hazards

    H226 - Flammable liquid and vapor

    EU Specific Hazard Statements

    EUH066 - Repeated exposure may cause skin dryness or cracking

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P403 + P233 - Store in a well-ventilated place. Keep container tightly closed