Thermo Scientific Chemicals

2-Amino-4-(Trifluormethyl)Thiophenolhydrochlorid, 97 %, Thermo Scientific Chemicals

Katalognummer: L09683.03
1 g, Each
Thermo Scientific Chemicals

2-Amino-4-(Trifluormethyl)Thiophenolhydrochlorid, 97 %, Thermo Scientific Chemicals

Katalognummer: L09683.03
1 g, Each
Menge
Katalognummer: L09683.03
auch als L09683-03 bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
4274-38-8
Appearance (Color)
White to cream to yellow or grey
Form
Crystalline powder or lumps
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
188-204°C

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute dermal toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Skin Sensitization
    Category 1
    Specific target organ toxicity - (repeated exposure)
    Category 2
    Acute aquatic toxicity
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H302 + H312 + H332 - Harmful if swallowed, in contact with skin or if inhaled

    H314 - Causes severe skin burns and eye damage

    H317 - May cause an allergic skin reaction

    H373 - May cause damage to organs through prolonged or repeated exposure

    H400 - Very toxic to aquatic life

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician