Thermo Scientific Chemicals

Diisopropyl-Bromomethylphosphonat, 97 %, Thermo Scientific Chemicals

Katalognummer: L13763.14
25 g, Each
Thermo Scientific Chemicals

Diisopropyl-Bromomethylphosphonat, 97 %, Thermo Scientific Chemicals

Katalognummer: L13763.14
25 g, Each
Menge
Katalognummer: L13763.14
auch als L13763-14 bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
98432-80-5
IUPAC Name
bis(propan-2-yl) (bromomethyl)phosphonate
Molecular Formula
C7H16BrO3P
InChI Key
JNONGIUWXHOEEN-UHFFFAOYSA-N
SMILES
CC(C)OP(=O)(CBr)OC(C)C
Form
Liquid or viscous liquid
Refractive Index
1.4485-1.4535 @ 20?C
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless

Beschreibung

Diisopropyl bromomethylphosphonate, is used as an organophosphorus intermediate. It is also used as a biochemical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Anwendungen
Diisopropyl-Bromomethyl-Phosphonat wird als Organophosphorintermediär verwendet. Wird auch als biochemisches Reagenz verwendet.

Löslichkeit
Nicht in Wasser löslich.

Hinweise
Behälter an einem trockenen und gut belüfteten Ort dicht verschlossen halten. Von Hitze und Oxidationsmitteln fernhalten. Stabil unter empfohlenen Lagerbedingungen.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H314 - Causes severe skin burns and eye damage

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician