Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: KSMQEVHIKT LEEIKLEKAL RVQQSSESST SSPSQHEATP GARRLLRITK RTGMKEEKNL QEGNEVDSQS SIRTEAKEAS GETTGVDITK IQVKRCETMR EKHMQKQQER EKSVLTPLRG DVASCNTQVA EKPVLTAVPG ITRHLTKRLP TKSSQKVEVE TSGIGDSLLN VKCAAQTLEK RGKAKPKVNV KPSVVKVVSS PKLAPKRKAV EMHAAVIAAV KPLSSSSVLQ EPPAKKAAVA VVPLVSEDKS VTVPEAENPR DSLVLPPTQS SSDSSPPEVS GPSSSQMSMK TRRLSSASTG KPPLSVEDDF EKLIWEISGG KLEAEIDLDP GKDEDDLLLE LSEMIDS
Involved in nuclear mRNA export; probably mediated by association with the TREX complex. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: unnamed protein product; Zinc finger CCCH domain-containing protein 11A; zinc finger CCCH-type domain containing 11A
Gene Aliases: KIAA0663; ZC3H11A; ZC3HDC11A
UniProt ID: (Human) O75152
Entrez Gene ID: (Human) 9877
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support