Compat-Able™ Protein Assay Preparation Reagent Kit
Thermo Scientific™

Compat-Able™ Protein Assay Preparation Reagent Kit

The Thermo Scientific Compat-Able Protein Assay Preparation Set eliminates interfering substances from protein assay samples to make them compatible withRead more
Have Questions?
Catalog NumberQuantity
23215500 mL
Catalog number 23215
Price (INR)
-
Quantity:
500 mL
Request bulk or custom format
The Thermo Scientific Compat-Able Protein Assay Preparation Set eliminates interfering substances from protein assay samples to make them compatible with BCA and Bradford assays.

The Compat-Able Reagents remove salts, detergents, reducing agents and other substances from protein samples to eliminate interference with protein assays. The protocol selectively precipitates all of the protein, thereby allowing the non-protein components to be easily decanted and removed. The purified, precipitated protein is redissolved in water, making it ready for accurate determination of the protein concentration by standard protein assay methods. The procedure is accomplished in less than 10 minutes with just a few simple steps. The Compat-Able Preparation Set is available individually or conveniently bundled with either the Thermo Scientific Pierce BCA Protein Assay Kit or the Thermo Scientific Pierce Coomassie Plus Protein Assay Kit.

Features of the Compat-Able Protein Assay Preparation Reagent Kit:

• Eliminate incompatibilities—removes nearly any substance that interferes with colorimetric protein assays: salts, detergents, denaturants, reducing agents, and even IEF sample buffer
• Simple and fast—prepare samples in less than 10 minutes with four simple steps
• Convenient—reagent set is easier to use than homemade TCA or acetone precipitation reagents, thereby producing more consistent results
• Scalable—microcentrifuge method is easily scaled to larger centrifuge tube volumes for special needs
• Validated—tested and offered together with BCA and Coomassie Plus Protein Assay Kits; protocol integrates control and normalization procedures

Related Products
Compat-Able™ BCA Protein Assay Kit
Compat-Able™ Coomassie Plus Protein Assay Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayBCA or Bradford Assay
DescriptionCompat-Able Protein Assay Preparation Reagent Set
For Use With (Application)Incompatibility Elimination
For Use With (Equipment)Spectrophotometer, Microplate Reader
Product LineCompat-Able
Product TypeProtein Quantitation Assay
Quantity500 mL
SpecificityNot Target-Specific
Detection MethodColorimetric
Unit SizeEach
Contents & Storage
Kit contains sufficient reagents to pretreat up to 500 samples
• 250 mL Compat-Able Protein Assay Preparation Reagent 1
• 250 mL Compat-Able Protein Assay Preparation Reagent 2

Store at room temperature