Product Image
Thermo Scientific™

Compat-Able™ BCA Protein Assay Kit

The Compat-Able BCA Protein Assay Kit eliminates interfering substances from protein assay samples to make them compatible with BCA andRead more
Have Questions?
Catalog NumberQuantity
232291 L
Catalog number 23229
Price (USD)
333.00
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
333.00
Each
The Compat-Able BCA Protein Assay Kit eliminates interfering substances from protein assay samples to make them compatible with BCA and Bradford assays.

See all available protein assays ›

The Compat-Able reagents included in this protein assay kit remove salts, detergents, reducing agents, and other substances from protein samples to eliminate interference with protein assays. The protocol selectively precipitates all of the protein, thereby allowing the non-protein components to be easily decanted and removed. The purified, precipitated protein is re-dissolved in water, making it ready for accurate determination of the protein concentration by standard protein assay methods. The procedure is accomplished in less than 10 minutes with just a few simple steps. The Compat-Able reagents are also available individually.

Features of the Compat-Able reagents include:
• Eliminate incompatibilities—removes nearly any substance that interferes with colorimetric protein assays: salts, detergents, denaturants, reducing agents, and even IEF sample buffer
• Simple and fast—prepare samples in less than 10 minutes with four simple steps
• Convenient—reagent set is easier to use than homemade TCA or acetone precipitation reagents, thereby producing more consistent results
• Scalable—microcentrifuge method is easily scaled to larger centrifuge tube volumes for special needs
• Validated—tested and offered together with BCA and Coomassie Plus Protein Assay Kits; protocol integrates control and normalization procedures
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayBCA Assay
DescriptionCompat-Able BCA Protein Assay Kit
For Use With (Application)Absorbance, Solution-based Detection, Incompatibility Elimination
For Use With (Equipment)Spectrophotometer, Microplate Reader
Product LineCompat-Able
Product TypeProtein Quantitation Assay
Quantity1 L
SpecificityNot Target-Specific
Sufficient For250 Tube Assays or 2500 Microplate Assays
Detection MethodColorimetric
Unit SizeEach
Contents & Storage
Sufficient For: Treating up to 500 protein assay samples and performing 250 tube assays or 2500 microplate assays
• Compat-Able Reagent 1, 250 mL
• Compat-Able Reagent 2, 250 mL
• BCA Reagent A, 500 mL
• BCA Reagent B, 25 mL
• Albumin Standard Ampules, 2 mg/mL, 10 x 1 mL

Frequently asked questions (FAQs)

What BCA protein assay kits do you recommend for NanoDrop 2000/2000c Spectrophotometers (Cat. Nos. ND-2000, ND-2000C)?

NanoDrop 2000/2000c Spectrophotometers (Cat. Nos. ND-2000, ND-2000C) are compatible with most commercial BCA protein assay kits. We offer a number of BCA protein assay kits, including, but not limited to, the BCA Protein Assay Kit (Cat. No. J63283.QA), Micro BCA Protein Assay Kit (Cat. No. 23235, and Pierce BCA Protein Assay Kit (Cat. No 23225, 23227).

To identify, compare, and choose the best protein assay for your sample type and application, you can use our interactive Protein Assay Selection Guide.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.