Thermo Scientific NHS-PEG4-Azide is an amine-reactive compound that can be used to derivatize primary amines of proteins or amine-coated polymerRead more
Have Questions?
Catalog Number
Quantity
26130
100 mg
Catalog number 26130
Price (INR)
27,403.00
線上優惠
Ends: 31-Dec-2026
45,671.00
Save 18,268.00 (40%)
Each
Quantity:
100 mg
Request bulk or custom format
Price (INR)
27,403.00
線上優惠
Ends: 31-Dec-2026
45,671.00
Save 18,268.00 (40%)
Each
Thermo Scientific NHS-PEG4-Azide is an amine-reactive compound that can be used to derivatize primary amines of proteins or amine-coated polymer surfaces for ligation to phosphine-modified molecules.
This NHS-ester compound reacts to form covalent bonds with primary amines (e.g., side chain of lysine residues or aminosilane-coated surfaces). The azide (N3) group reacts with phosphine-labeled molecules by a mechanism known as Staudinger chemistry, enabling efficient and specific conjugation of derivatized molecules in biological samples.
Features of NHS-PEG4-Azide:
• Soluble—reagent easily dissolves in water-miscible solvents for subsequent dilution in aqueous reaction mixtures with cell lysates and other biological samples • Compatible—reaction chemistry occurs effectively in simple buffer conditions; requires no accessory reagents such as copper or reducing agents • Specific—NHS esters are specific for covalent attachment to primary amines (side chain of lysine or N-terminus of polypeptides) • Chemoselective—azide and phosphine groups do not react or interfere with components of biological samples (bioorthogonal) but conjugate to one another with high efficiency
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityNot determined
DescriptionNHS-PEG4-Azide
FormLiquid
Labeling MethodChemical Labeling
Molecular Weight (g/mol)388.37
PEGylatedYes
Product LinePierce
Quantity100 mg
Reactive MoietyAmine, Azide, NHS Ester
Shipping ConditionAmbient
SolubilityDMF, DMSO
Spacer Arm Length18.9 Å
Water SolubleNo
Chemical ReactivityAmine-Phosphine
CleavableNo
Crosslinker TypeHeterobifunctional
FormatStandard
Product TypeCrosslinker
SpacerMedium (10 to 30 Å)
Unit SizeEach
Contents & Storage
Upon receipt store at -20°C protected from light and moisture.
Frequently asked questions (FAQs)
Can you provide the shelf-life for NHS-PEG4-Azide?
NHS-PEG4-Azide is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (-20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.