NHS-Fluorescein (5/6-carboxyfluorescein succinimidyl ester), mixed isomer
Thermo Scientific™

NHS-Fluorescein (5/6-carboxyfluorescein succinimidyl ester), mixed isomer

NHS-Fluorescein is an amine-reactive derivative of fluorescein dye that has wide-ranging applications as a label for antibodies and other probes,Read more
Have Questions?
Catalog NumberQuantity
464091 g
Catalog number 46409
Price (INR)
-
Quantity:
1 g
Request bulk or custom format
NHS-Fluorescein is an amine-reactive derivative of fluorescein dye that has wide-ranging applications as a label for antibodies and other probes, for use in fluorescence microscopy, flow cytometry and immunofluorescence-based assays such as Western blotting and ELISA. The NHS-ester of fluorescein efficiently labels antibodies and other purified proteins at primary amines (lysine side chains)

Properties of NHS-Fluorescein:

• Alternative name: 5/6-FAM SE
• Chemical name: 5/6-carboxyfluorescein succinimidyl ester
• Molecular weight: 473.4
• Excitation source: 488 nm spectral line, argon-ion laser
• Excitation wavelength: 494 nm
• Emission wavelength: 518 nm
• Extinction coefficient: > 70,000 M-1cm-1
• CAS #: 117548-22-8
• Purity: > 90% by HPLC
• Solubility: Soluble in DMF or DMSO
• Reactive groups: NHS ester, reacts with primary amines at pH 7.0 to 9.0

NHS-fluorescein is activated with the N-hydroxy-succinimidyl-ester (NHS ester) functional group. Compared to FITC, the NHS-ester deriviative has greater specificity toward primary amines in the presence of other nucleophiles and results in a more stable linkage following labeling. Pierce Amine-reactive Fluorescein Dyes are mixtures of isomers with reactive groups attached at the 5- and 6-positions of the bottom ring. The properties of these isomers are indistinguishable in terms of excitation and emission spectra, and for protein applications there is no need to isolate a specific isomer.

Applications
• Label antibodies for use as immunofluorescent probes
• Label oligonucleotides for hybridization probes
• Detect proteins in gels and on Western blots

Related Products
FITC (5/6-fluorescein isothiocyanate), mixed isomer
Pierce™ NHS-Fluorescein Antibody Labeling Kit
Pierce™ FITC Antibody Labeling Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAmine
Emission518
Excitation494
Label or DyeFITC (Fluorescein)
Product TypeNHS-Fluorescein
Quantity1 g
Reactive MoietyActive Ester, Succinimidyl Ester, NHS Ester
SolubilityDMF (Dimethylformamide)
Label TypeClassic Dyes
Product LinePierce
Unit SizeEach
Contents & Storage
Store desiccated at -20°C, protect from moisture

Frequently asked questions (FAQs)

Can you provide the shelf-life for NHS-Fluorescein (5/6-carboxyfluorescein succinimidyl ester), mixed isomer?

NHS-Fluorescein (5/6-carboxyfluorescein succinimidyl ester), mixed isomer is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (-20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.