FITC (5/6-fluorescein isothiocyanate), mixed isomer
Actual product may vary
FITC (5/6-fluorescein isothiocyanate), mixed isomer
FITC (5/6-fluorescein isothiocyanate), mixed isomer
Thermo Scientific™

FITC (5/6-fluorescein isothiocyanate), mixed isomer

Fluorescein isothiocyanate (FITC) is an amine-reactive derivative of fluorescein dye that has wide-ranging applications as a label for antibodies andRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
46425100 mg
464241 g
Catalog number 46425
Price (INR)
9,209.00
Online offer
Ends: 31-Dec-2026
15,347.00
Save 6,138.00 (40%)
Each
Quantity:
100 mg
Request bulk or custom format
Price (INR)
9,209.00
Online offer
Ends: 31-Dec-2026
15,347.00
Save 6,138.00 (40%)
Each
Fluorescein isothiocyanate (FITC) is an amine-reactive derivative of fluorescein dye that has wide-ranging applications as a label for antibodies and other probes, for use in fluorescence microscopy, flow cytometry and immunofluorescence-based assays such as Western blotting and ELISA. The isothiocyanate variety of fluorescein efficiently labels antibodies and other purified proteins at primary amines (lysine side chains)

Properties of FITC:

• Alternative names: 5/6-FITC
• Chemical name: 5(6)-fluorescein isothiocyanate mixed isomer
• Molecular weight: 389.2
• Excitation source: 488 nm spectral line, argon-ion laser
• Excitation wavelength: 494 nm
• Emission wavelength: 518 nm
• Extinction coefficient: > 70,000 M-1cm-1
• CAS #: 27072-45-3
• Purity: > 95% by HPLC
• Solubility: Soluble in aqueous buffers at pH > 6
• Reactive groups: Isothiocyanate, reacts with primary amines at pH 7.0 to 9.0

Applications
• Label antibodies for use as immunofluorescent probes
• Label oligonucleotides for hybridization probes
• Detect proteins in gels and on Western blots

Related Products
NHS-Fluorescein (5/6-carboxyfluorescein succinimidyl ester), mixed isomer
Pierce™ NHS-Fluorescein Antibody Labeling Kit
Pierce™ FITC Antibody Labeling Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAmine
Emission518
Excitation494
Label or DyeFITC (Fluorescein)
Product TypeFITC
Quantity100 mg
Reactive MoietyIsothiocyanate
SolubilityDMF (Dimethylformamide)
Label TypeClassic Dyes
Product LinePierce
Unit SizeEach
Contents & Storage
Store desiccated at -20°C, protect from moisture