B-27™ Supplement, minus insulin
Gibco™

B-27™ Supplement, minus insulin

A variation of the most cited neuronal cell culture supplement, B-27 Supplement Minus Insulin is a customized version of theRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A1895602100 mL
A189560110 mL
Catalog number A1895602
Price (INR)
-
Quantity:
100 mL
A variation of the most cited neuronal cell culture supplement, B-27 Supplement Minus Insulin is a customized version of the original B-27 Supplement without insulin. B-27 Supplement Minus Insulin is a serum-free supplement designed to support growth of hippocampal and other CNS neurons in applications where the presence of insulin causes interference. B-27 Supplement Minus Insulin is provided as a 50X liquid and is intended for use with Neurobasal Medium or Neurobasal-A Medium.

B-27 Supplement is suitable for use in most neuronal cell applications. For other applications, we provide alternative formulations of B-27 Supplement and Neurobasal media.

For established protocols using B-27 Supplement, access our Gibco Neurobiology Protocol Handbook.

View publications using Gibco neurobiology products at our Gibco CellCite.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeNeural Cells
Concentration50 X
Culture TypeMammalian Cell Culture
Product LineB-27
Quantity100 mL
Serum LevelSerum-Free
Shelf Life12 Months
Shipping ConditionDry Ice
SourceSerum-Free
Tested ForEndotoxin, Performance, Sterility, Mycoplasma
ClassificationSerum-free
FormLiquid
Product TypeCell Culture Supplement
SterilitySterile
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -20°C) and protect from light.

Frequently asked questions (FAQs)

Do you offer B-27 supplements with and without insulin?

Yes, we do offer B-27 Supplement, minus insulin (Cat. Nos. A1895601 and A1895602). All our other B-27 supplements do contain insulin.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.