Sulfo-NHS (N-hydroxysulfosuccinimide), No-Weigh™ Format
Thermo Scientific™

Sulfo-NHS (N-hydroxysulfosuccinimide), No-Weigh™ Format

Thermo Scientific Pierce Sulfo-NHS, No-Weigh Format is a chemical modification reagent for converting carboxyl groups to amine-reactive NHS esters forRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A3926910 x 2 mg
24510500 mg
245255 g
Catalog number A39269
Price (INR)
-
Quantity:
10 x 2 mg
Thermo Scientific Pierce Sulfo-NHS, No-Weigh Format is a chemical modification reagent for converting carboxyl groups to amine-reactive NHS esters for bioconjugation, crosslinking, labeling and immobilization methods.

Features of Sulfo-NHS:

• Efficiency of EDC-mediated coupling is increased in the presence of Sulfo-NHS
• Amine-reactive NHS esters or Sulfo-NHS esters can be made with any carboxyl-containing molecule
• Sulfo-NHS derivatives are usually directly water-soluble (can be added directly to physiologic buffers) and membrane-impermable (can be used for cell surface labeling)
• High-purity, crystalline Sulfo-NHS can be used to create high-quality activated derivatives

Sulfo-NHS (N-hydroxysulfosuccinimide) enables control and modification of carbodiimide crosslinking reactions involving activation of carboxylates (—COOH) for conjugation with primary amines (—NH2). Derivatives are easily synthesized by mixing the Sulfo-NHS with a carboxyl-containing molecule and a dehydrating agent such as the carbodiimide EDC (EDAC). The method is the basis for generating many types of protein labeling reagents, including amine-reactive fluorescent dyes, biotin affinity tags and pegylation compounds.

Applications:
• Improve efficiency of EDC coupling reactions
• Convert carboxyls to amine-reactive Sulfo-NHS esters
• Crosslink proteins to carboxyl-coated beads or surfaces more efficiently
• Activate nanoparticles with amine-reactive Sulfo-NHS esters

Specifications for Sulfo-NHS:
We manufacture N-hydroxysulfosuccinimide to the highest possible specifications to produce the most specific bioconjugates, to ensure the integrity of your data and to provide you with the highest degree of consistency. Each lot of Sulfo-NHS is tested to meet the following minimum specifications:

• Purity—Greater than 95% by quantitative NMR (the highest standard for crosslinker purity);
average lot purity is greater than 99%
• Solubility—Sample dissolves at 2 mg/mL in deionized water to yield a clear, colorless solution
• Identity—IR scan shows only peaks characteristic of N-hydroxysulfosuccinimide

Product References:
Crosslinker Application Guide -- search for recent literature references for this product

Related Products
Pierce™ Premium Grade Sulfo-NHS (N-hydroxysulfosuccinimide)
NHS (N-hydroxysuccinimide)
EDC (1-ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityNo
DescriptionSulfo-NHS
FormSolid
Labeling MethodChemical Labeling
Molecular Weight (g/mol)217.13
PEGylatedNo
Product LineNo-Weigh
Quantity10 x 2 mg
Reactive MoietySulfo-NHS Ester
Shipping ConditionAmbient
SolubilityWater
Spacer Arm LengthShort
Water SolubleYes
Chemical ReactivityAmine-Carboxyl
CleavableNo
Crosslinker TypeHeterobifunctional
FormatStandard, Single-use, Premium-grade
Product TypeCrosslinker
SpacerShort (<10 Å)
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C.

Frequently asked questions (FAQs)

Can you provide the shelf-life for Sulfo-NHS (N-hydroxysulfosuccinimide)?

Sulfo-NHS (N-hydroxysulfosuccinimide) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (4 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Can you provide the shelf life for Sulfo-NHS (N-hydroxysulfosuccinimide)?

Sulfo-NHS is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

How does a product with "No-Weigh" format reduce my concern over reagent stability?

The No-Weigh product format, where small amounts of reagent are pre-weighed and packaged separately, prevents the contamination and loss of reagent reactivity over time associated with repetitive opening and closing of a single vial. This format enables use of a fresh vial of reagent each time and eliminates the hassle of weighing out small amounts of reagent. Please view additional information and the video at: https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-labeling-crosslinking/no-weigh-packaging.html

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.