SuperSignal™ Western Blot Substrate Bundle, Femto + trial-size Pico PLUS
Thermo Scientific™

SuperSignal™ Western Blot Substrate Bundle, Femto + trial-size Pico PLUS

This substrate kit is comprised of SuperSignal West Femto Maximum Sensitivity Substrate plus a sample size of SuperSignal West PicoRead more
Have Questions?
Catalog NumberQuantity
A43841200 mL bundle
Catalog number A43841
Price (INR)
-
Quantity:
200 mL bundle
This substrate kit is comprised of SuperSignal West Femto Maximum Sensitivity Substrate plus a sample size of SuperSignal West Pico PLUS Chemiluminescent Substrate to provide chemiluminescent detection substrates for both high sensitivity and routine western blotting.

SuperSignal West Femto Maximum Sensitivity Substrate
Thermo Scientific SuperSignal West Femto Maximum Sensitivity Substrate is an ultra-sensitive enhanced chemiluminescent (ECL) substrate for low femtogram protein level detection by western blot analysis with horseradish peroxidase (HRP) enzyme. It is optimized to detect low abundant and/or difficult to detect targets.

• Sensitivity: low-femtogram (mid-zeptomole)
• Stability: 8-hr working solution stability, 1-year kit stability at 4°C
• Compatibility: nitrocellulose and PVDF membranes
• Signal duration: up to 8 hours
• Recommended primary antibody concentration: 1:5,000–1:100,000 dilution (10–200 ng/mL)
• Recommended secondary antibody concentration: 1:100,000–1:500,000 dilution (2–10 ng/mL)

View full product information ›

SuperSignal West Pico PLUS Chemiluminescent Substrate
Thermo Scientific SuperSignal West Pico PLUS Chemiluminescent Substrate enables picogram to high femtogram-level protein detection by western blot analysis. It is best for routine, everyday western blotting applications, providing excellent signal intensity, sensitivity and duration across a wide range of targets.

• Sensitivity: pictogram to femtogram sensitivity
• Stability: 8-hr working solution stability; 1-year kit stability at room temperature
• Compatibility: nitrocellulose and PVDF membranes
• Signal duration: 6 to 24 hours
• Recommended primary antibody concentration: 1:1,000–1:5,000 dilution (0.2–1.0 μg/mL)
• Recommended secondary antibody concentration: 1:20,000–100,000 dilution (10–50 ng/mL)

View full product information ›

For Research Use Only. Not for use in diagnostic procedures.
Specifications
SensitivityPicogram to femtogram, low femtogram (mid-zeptomole)
Shipping ConditionRoom Temperature
Sufficient For60 Mini-Gel Blots, 5000 sq. cm of membrane; 2 Mini-Gel Blots, 200 sq. cm of Membrane
Detection MethodChemiluminescence
For Use With (Application)Western Blot
FormLiquid
Membrane CompatibilityNitrocellulose, PVDF
Product LineSuperSignal
Product TypeSubstrate
Quantity200 mL bundle
SubstrateHorseradish Peroxidase
Unit SizeEach
Contents & Storage
• 200 mL SuperSgnal West Femto reagent, store at 2–8°C
• 20 mL SuperSignal West Pico PLUS reagent, store at room temperature