Product Image
Invitrogen™
iBright™ SAE Software for 21 CFR Part 11, single license
iBright SAE Software for 21 CFR Part 11 is for use with the iBright Imaging System and supports compliance withRead more
Have Questions?
Catalog NumberQuantity
A492081 license
Catalog number A49208
Price (INR)
-
Quantity:
1 license
iBright SAE Software for 21 CFR Part 11 is for use with the iBright Imaging System and supports compliance with 21 CFR Part 11 FDA guidelines for security, auditing, and electronic (SAE) signatures using the SAE console.
Features of the iBright SAE Software include:
Security
• Authentication of user login information
• Ability to set password guidelines and expiry dates
• Lockout access and re-authentication requirements after a period of inactivity
• System lockout upon unauthorized attempts to access software
• Ability of administrator to define user roles and permissions
• Verification of each user's permissions to perform predefined activities
• Identification of data tampering
Auditing
• Generation of audit trail log
• Tracking of changes and committed actions to user roles, account management, and system settings
• Ability of administrator to determine functions required to input reason for commit
• Audit trail record presented in readable and filterable formats
Electronic signature
• Administrator determination of e-signature requirements
• Ability to review records and sign electronically
Along with the 21 CFR part 11 software, our service and support organization offers IQ/OQ qualification services for your instrument and support training including SmartStart services.
Specifications
Quantity1 license
Software TypeSoftware License
For Use With (Equipment)iBright Imaging System
Product LineiBright
TypeSoftware License
Unit SizeEach
Frequently asked questions (FAQs)
Are the iBright Imaging Systems network-compatible?
The iBright Imaging Systems support Ethernet network connections. WIFI is also supported with the addition of a USB-WIFI dongle. Data may be exported from the instrument via network and WIFI connections. It can also be exported via USB port.
For additional information on iBright functionality, visit this page.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Is a computer needed to control the iBright Imaging Systems?
iBright Imaging Systems feature a built-in computer interface controlled by the 12.1 inch touchscreen. No additional computer is needed to operate the instrument.
For additional information on iBright functionality, visit this page.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Do the iBright Imaging Systems support .tiff file export?
iBright imagers support the export of .tiff files in 8 and 16 bit formats. They also allow users to export images as .jpg and the proprietary .g2i file types. The iBright Analysis software primarily uses .g2i files as this format contains the most data to facilitate easy analysis. But it also supports .tiff files import and analysis.
For additional information on iBright functionality, visit this page.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Is analysis software included with purchase of an iBright Imaging System?
The free iBright Analysis Software (iBA) supports analysis of all images captured on iBright imagers and is available for free download on our website. There is also a Cloud version of iBA software available for users who prefer to work directly from a web browser.
The iBA software is supported on both Windows and Mac computers and can be used by any lab personnel who need it. This software automates data analysis and makes annotation, analysis, and reporting simple. To learn more about the iBA software, view the iBright Analysis Software how-to-videos available on this page.
To speak with a representative about the iBright imager or request a quote or demo, please fill out this form.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
What are the primary differences between the iBright SAE secure version of the iBright Analysis software and the standard version of the software?
Most functionality is the same between the standard and secure versions of the iBright Analysis (iBA) software. The primary difference is that the secure version of the software supports the auditing and signature functionality that is needed to support 21CFR laboratories.
To learn more about iBA software, visit this page.
To speak with a representative about the iBright imager or request a quote or demo, please fill out this form.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.