Flat PCR Caps, strips of 12
Thermo Scientific™

Flat PCR Caps, strips of 12

The Thermo Scientific Flat PCR Cap ,Strip of 12 are used for PCR applications and work with the Thermo ScientificRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorProduct Type
AB0850NaturalFlat Cap, 12-Cap Strip
AB0851NaturalPCR Plate Cap Strip
Catalog number AB0850
Price (INR)
5,666.00
Offre exceptionnelle en ligne
Ends: 31-Dec-2026
9,442.00
Save 3,776.00 (40%)
Each
Color:
Natural
Product Type:
Flat Cap, 12-Cap Strip
Request bulk or custom format
Price (INR)
5,666.00
Offre exceptionnelle en ligne
Ends: 31-Dec-2026
9,442.00
Save 3,776.00 (40%)
Each
The Thermo Scientific Flat PCR Cap ,Strip of 12 are used for PCR applications and work with the Thermo Scientific 96-well PCR plates and strip tubes. These cap strips are available in either 8- or 12-cap strips. The Flat PCR Cap Strips signature flat cap allows you to label each sample.

Features of the Thermo Scientific Flat PCR Cap Strip

• 8- and 12-cap strips
• Suitable for use with 96-well PCR plates as well as standard and low profile strips
• Flat surface of each cap facilitates sample labeling
• 25 packs of 8

Related Products
Flat PCR Caps, strips of 8
Flat PCR Caps, strips of 12
Low Profile Tubes, strips of 12
Tubes, strips of 12
Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Equipment)Thermal Cycler
Format80 Strip Caps
Product TypeFlat Cap, 12-Cap Strip
Quantity80 strips
ColorNatural
MaterialPolypropylene
Unit SizeEach