A more sustainable equivalent is available.
Domed and Flat PCR Cap Strips
Thermo Scientific™
Domed and Flat PCR Cap Strips
The Thermo Scientific Flat PCR Cap ,Strip of 8 are used for PCR applications and work with the Thermo ScientificRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityColor
AB0783120/Cs.Clear
AB0784250/Cs.Clear
Catalog number AB0783
Price (USD)
52.65
Online exclusive
62.50
Save 9.85
Each
Quantity:
120/Cs.
Color:
Clear
Request bulk or custom format
Price (USD)
52.65
Online exclusive
62.50
Save 9.85
Each
The Thermo Scientific Flat PCR Cap ,Strip of 8 are used for PCR applications and work with the Thermo Scientific 96-well PCR plates and strip tubes. These cap strips are available in either 8- or 12-cap strips. The Flat PCR Cap Strips signature flat cap allows you to label each sample.
Features of the Thermo Scientific Flat PCR Cap Strip
• 8- and 12-cap strips
• Suitable for use with 96-well PCR plates as well as standard and low profile strips
• Flat surface of each cap facilitates sample labeling
Related Products
Flat PCR Caps, strips of 8
Flat PCR Caps, strips of 12
Specifications
FormatStrips of 8 Flat Caps
Product TypePCR Plate Cap Strip
Quantity120/Cs.
Certified RNase/DNase-freeYes
ColorClear
MaterialPolypropylene
No. of Wells8
Plate Compatibility96-well Plate
Unit SizeEach