VersiPlate PCR Strip Tube Plate, 96-well, low profile, white
Thermo Scientific™

VersiPlate PCR Strip Tube Plate, 96-well, low profile, white

The Thermo Scientific VersiPlate, White 96-well Low Profile PCR Strip Tube Plate, is unique, with each strip of eight tubesRead more
Have Questions?
Catalog NumberQuantity
AB1800W25 Plates
Catalog number AB1800W
Price (INR)
16,297.00
Each
Quantity:
25 Plates
Price (INR)
16,297.00
Each
The Thermo Scientific VersiPlate, White 96-well Low Profile PCR Strip Tube Plate, is unique, with each strip of eight tubes being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.

Features

• Easier sample handling as multiple strips of samples can be handled simultaneously
• Reduced consumables waste as only the required numbers of strips need to be removed with the remainder being stored for the next experiment
• Can be used in instruments with or without a VersiFrame.
• The reusable polycarbonate frame can serve as a useful rack for a VersiPlate or any number of low profile strip tubes and facilitates sample loading into thermal cyclers
• The low profile format of the VersiPlate has a lower dead volume which reduces sample evaporation
• Available in white which improves signal reflection, reduces variation and increases the sensitivity of qPCR assays
• Maximum fill volume of 0.2 mL
• Can be used with all Thermo Scientific VersiCap™ Mats, cap strips, adhesive and heat seals

Applications

• qPCR/Real-Time PCR

Related Products
VersiPlate PCR Strip Tube Plate, 96-well, low profile
VersiPlate Frame, 96-well, skirted, whiteVersiCap Mat, 96-well, domed cap strips
VersiCap Mat, 96-well, flat cap strips
VersiCap Mat, 96-well, ultra clear cap strips
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging25 Plate
Product TypePCR Microplate
Quantity25 Plates
Reaction SpeedLow profile
Volume (Metric) Working0.2 mL
BarcodeNo Barcode
ColorWhite
MaterialPolycarbonate
No. of Wells96
Product LineVersiPlate
Volume (Metric) Well0.2 mL
Unit SizeEach

Frequently asked questions (FAQs)

Why should I use Thermo Scientific white plastics in qPCR?

White plates and tubes reflect significantly more signal back to the qPCR instrument's detector than traditional clear plates. The improved signal reflection ensures that even the lowest levels of fluorescence are detected. White well walls also prevent signal from passing through to the cycler block where it can be inconsistently reflected or absorbed. This keeps variations in the cycler block from affecting qPCR data. Using Thermo Scientific white PCR plastics increases sensitivity and reduces variability within the qPCR assay.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.