E-Gel™ Power Snap Electrophoresis Device Starter Kit, DC Safe 1%
Invitrogen™
E-Gel™ Power Snap Electrophoresis Device Starter Kit, DC Safe 1%
The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit, DC Safe 1%, provides everything you need to quickly start usingRead more
Have Questions?
Catalog NumberQuantity
G8171ST1 kit
Catalog number G8171ST
Price (INR)
-
Quantity:
1 kit
The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit, DC Safe 1%, provides everything you need to quickly start using precast E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain on the device.
The kit includes:
  • Ten E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 1% (Cat. No. A42347)
  • E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
  • E-Gel 1-Kb Plus Express DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488091)
Learn more about the E-Gel Power Snap Electrophoresis System ›
Learn more about precast E-Gel EX agarose gel technology ›
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 kit
TypeElectrophoresis System Starter Kit
Unit SizeEach
Contents & Storage
• Ten E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 1% (Cat. No. A42347)
• E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
• E-Gel 1-Kb Plus Express DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488091)
Store at room temperature.