Product Image

Productainer™ BioProcess Container (BPC), 50 L with 2 Top and 1 Bottom Ports, 3D Single-Use BPC

Have Questions?
Catalog number SH3067201
Price (INR)
-
The Thermo Scientific™ Productainer™ BioProcess Container (BPC), 50 L 3D system is composed of CX5-14 single web, multi-layer plastic film and includes 2 top ports and 1 bottom port. The 3D square tube design with bottom port dispense is for use with conical, bottom-drain drums.

Tubing style:
• Line 1: 19.1mm (.75 in.) tri-clamp, polyethylene, Tubing: length: 45.7cm (18 in.) ID x OD: 12.7mm x 19.1mm (.5 in. x .75 in.)
• Line 2: 19.1mm (.75 in.) tri-clamp, polyethylene, Tubing: length: 45.7cm (18 in.) ID x OD: 12.7mm x 19.1mm (.5 in. x .75 in.)
• Line 3: 19.1mm (.75 in.) tri-clamp, polyethylene, Tubing: C-Flex, length: 121.9cm (48 in.) ID x OD: 12.7mm x 19.1mm (.5 in x .75 in.)

Features:
• Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
• All 3D BPCs are constructed in an ISO 7 cleanroom under cGMP conditions.
• All 3D BPCs are designed to fit the full range of support containers, both square and cylindrical – from 50 to 2000 L.

Applications of the Thermo Scientific 3D BPC include:
• Hydration and sterilization of process buffers, liquids, and culture media.
• Chromatography feed and fraction collection.
• Storage and transport of bulk drug product and bulk drug precursors.
• Harvest from and feed into bioreactors and fermentors.
• Dispensing, packaging, and storage of cell culture media, buffers and process liquids.

Customize standard BPC systems for optimized single-use technology. Choose from the industry’s largest component library – with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
• Filters—used for bioburden reduction; sterilization grade filters and vent filters.
• Tubing—thermoplastic elastomers, platinum-cured silicone.
• Sample ports—septum-style and needle-free ports.
• Clamps—sliders, pinch, and crimp bands.

Related Products:
Productainer™ BioProcess Container (BPC), 50L with 2 Top Ports, 3D Single-Use
Productainer™ BioProcess Container (BPC), 100L with 2 Top Ports, 3D Single-Use
Productainer™ BioProcess Container (BPC), 200L with 2 Top Ports, 3D Single-Use

Productainer™ BioProcess Container (BPC), 50L with 2 Top and 1 Bottom Ports, 3D Single-Use
Productainer™ BioProcess Container (BPC), 100L with 2 Top and 1 Bottom Ports, 3D Single-Use
Productainer™ BioProcess Container (BPC), 200L with 2 Top and 1 Bottom Ports, 3D Single-Use

Productainer™ BioProcess Container (BPC), 50 L with 4 Top Ports, 3D Single-Use
Productainer™ BioProcess Container (BPC), 100 L with 4 Top Ports, 3D Single-Use
Productainer™ BioProcess Container (BPC), 200 L with 4 Top Ports, 3D Single-Use

Productainer™ BioProcess Container (BPC), 50 L with 3 Top and 1 Bottom Ports, 3D Single-Use
Productainer™ BioProcess Container (BPC), 100 L with 4 TopPorts and 1 Bottom Dispense, 3D Single-Use
Productainer™ BioProcess Container (BPC), 200 L with 4 TopPorts and 1 Bottom Dispense, 3D Single-Use

Productainer™ BioProcess Container (BPC), 50 L with 4 Top Drains, 3D Single-Use
Productainer™ BioProcess Container (BPC), 100 L with 4 Top Drains, 3D Single-Use
Productainer™ BioProcess Container (BPC), 200 L with 4 Top Drains, 3D Single-Use

Productainer™ BioProcess Container (BPC), 50 L with 2 Top and 1 Bottom Ports, 3D Single-Use
Productainer™ BioProcess Container (BPC), 100 L with 2 Top and 1 Bottom Ports, 3D Single-Use
Productainer™ BioProcess Container (BPC), 200 L with 2 Top and 1 Bottom Ports, 3D Single-Use

Specifications
Unit Size50 L