Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 241009_mat
          Assay Name: hsa-miR-34c-3p
          miRBase Accession Number: MI0000743
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 111513439 - 111513515 [+] on Build GRCh38
          Mature miRNA Sequence: AAUCACUAACCACACGGCCAGG
          Species: Human, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 241009_mat
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000039, mir-34

          Mature miRNA Sequence:

          AAUCACUAACCACACGGCCAGG

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-34c-3p MIMAT0004677
          Cricetulus griseus cgr-miR-34c-3p MIMAT0023930
          Dasypus novemcinctus dno-miR-34c-3p MIMAT0047580
          Goat chi-miR-34c-3p MIMAT0036170
          Guinea pig cpo-miR-34c-3p MIMAT0046927
          Pan troglodytes ptr-miR-34c MIMAT0008114
          View More Rows
          Pongo pygmaeus ppy-miR-34c-3p MIMAT0015743
          Pteropus alecto pal-miR-34c-3p MIMAT0039792
          Rabbit ocu-miR-34c-3p MIMAT0048172
          Rhesus monkey mml-miR-34c-3p MIMAT0006176

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-34c
          Stem-loop Accession # MI0000743
          Stem-loop Sequence
          AGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGGUAAAAAGAUU
          Chromosome Location Chr. 11 - 111513439 - 111513515 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004677
          miRBase ID: hsa-miR-34c-3p
          Mature miRNA Sequence: AAUCACUAACCACACGGCCAGG
          Chromosome Location: Chr. 11 - 111513439 - 111513515 [+] on Build GRCh38

          Cricetulus griseus

          Stem-loop ID cgr-mir-34c
          Stem-loop Accession # MI0020507
          Stem-loop Sequence
          CUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGGUAAAAAGACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023930
          miRBase ID: cgr-miR-34c-3p
          Mature miRNA Sequence: AAUCACUAACCACACGGCCAGG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-34c
          Stem-loop Accession # MI0038946
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047580
          miRBase ID: dno-miR-34c-3p
          Mature miRNA Sequence: AAUCACUAACCACACGGCCAGG
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-34c
          Stem-loop Accession # MI0030759
          Stem-loop Sequence
          UGAGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAAUACCAAUCACUAACCACACGGCCAGGUAAAAAGAUUUGGGAAUUCAUCCAAAUGAGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036170
          miRBase ID: chi-miR-34c-3p
          Mature miRNA Sequence: AAUCACUAACCACACGGCCAGG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-34c
          Stem-loop Accession # MI0038602
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046927
          miRBase ID: cpo-miR-34c-3p
          Mature miRNA Sequence: AAUCACUAACCACACGGCCAGG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-34c
          Stem-loop Accession # MI0008633
          Stem-loop Sequence
          GUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGGUAAAAAGAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008114
          miRBase ID: ptr-miR-34c
          Mature miRNA Sequence: AAUCACUAACCACACGGCCAGG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-34c
          Stem-loop Accession # MI0014808
          Stem-loop Sequence
          AGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGGUAAAAAGAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015743
          miRBase ID: ppy-miR-34c-3p
          Mature miRNA Sequence: AAUCACUAACCACACGGCCAGG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-34c
          Stem-loop Accession # MI0032331
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039792
          miRBase ID: pal-miR-34c-3p
          Mature miRNA Sequence: AAUCACUAACCACACGGCCAGG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-34c
          Stem-loop Accession # MI0039258
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGCUGAUAGUACCAAUCACUAACCACACGGCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048172
          miRBase ID: ocu-miR-34c-3p
          Mature miRNA Sequence: AAUCACUAACCACACGGCCAGG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-34c
          Stem-loop Accession # MI0007605
          Stem-loop Sequence
          AGUCUAGUUACCAGGCAGUGUAGUUAGCUGAUUGCUGAUAGUACCAAUCACUAACCACACGGCCAGGUAAAAAGAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006176
          miRBase ID: mml-miR-34c-3p
          Mature miRNA Sequence: AAUCACUAACCACACGGCCAGG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03295169_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12342
          Ambion® Pre-miR™ miRNA Precursor : PM12342
          mirVana® miRNA inhibitor : MH12342
          mirVana® miRNA mimic : MC12342
          TaqMan™ Advanced miRNA Assay : 478051_mir

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline