Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 462557_mat
          Assay Name: rno-miR-34a*
          miRBase Accession Number: MI0000877
          miRBase Version: v22.1
          Chromosome Location: Chr.5: 167092491 - 167092592 [+] on Build Rnor_6.0
          Mature miRNA Sequence: AAUCAGCAAGUAUACUGCCCUA
          Species: Rat, Chrysemys picta, Dasypus novemcinctus, Guinea pig, Monodelphis domestica, Rabbit, Rhesus monkey
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 462557_mat
          Size
          Availability Made To Order
          Catalog # 4440886
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000039, mir-34

          Mature miRNA Sequence:

          AAUCAGCAAGUAUACUGCCCUA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-34a-3p MIMAT0017106
          Chrysemys picta cpi-miR-34a-3p MIMAT0037693
          Dasypus novemcinctus dno-miR-34a-3p MIMAT0047576
          Guinea pig cpo-miR-34a-3p MIMAT0046923
          Monodelphis domestica mdo-miR-34a-3p MIMAT0026655
          Rabbit ocu-miR-34a-3p MIMAT0048168
          View More Rows
          Rhesus monkey mml-miR-34a-3p MIMAT0026586

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-34a
          Stem-loop Accession # MI0000877
          Stem-loop Sequence
          CCGGCUGUGAGUAAUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGUAUUAGCUAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUGCACGUUGU
          Chromosome Location Chr. 5 - 167092491 - 167092592 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017106
          miRBase ID: rno-miR-34a-3p
          Mature miRNA Sequence: AAUCAGCAAGUAUACUGCCCUA
          Chromosome Location: Chr. 5 - 167092491 - 167092592 [+] on Build Rnor_6.0

          Chrysemys picta

          Stem-loop ID cpi-mir-34a
          Stem-loop Accession # MI0029434
          Stem-loop Sequence
          GUGAGUGUUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGCAAUAAUCAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUGCACAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037693
          miRBase ID: cpi-miR-34a-3p
          Mature miRNA Sequence: AAUCAGCAAGUAUACUGCCCUA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-34a
          Stem-loop Accession # MI0038944
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGAGUAAUAGUGCAGGAAGCAAUCAGCAAGUAUACUGCCCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047576
          miRBase ID: dno-miR-34a-3p
          Mature miRNA Sequence: AAUCAGCAAGUAUACUGCCCUA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-34a
          Stem-loop Accession # MI0038600
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGAGUAUUGGUGCAGGAAGCAAUCAGCAAGUAUACUGCCCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046923
          miRBase ID: cpo-miR-34a-3p
          Mature miRNA Sequence: AAUCAGCAAGUAUACUGCCCUA
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-34a
          Stem-loop Accession # MI0005280
          Stem-loop Sequence
          GGCCAGCUGUGAGUGUUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGUAAUAGAUAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUGCACGUUGUUAGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026655
          miRBase ID: mdo-miR-34a-3p
          Mature miRNA Sequence: AAUCAGCAAGUAUACUGCCCUA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-34a
          Stem-loop Accession # MI0039256
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGAGUCACAGAGAAGGAAGCAAUCAGCAAGUAUACUGCCCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048168
          miRBase ID: ocu-miR-34a-3p
          Mature miRNA Sequence: AAUCAGCAAGUAUACUGCCCUA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-34a
          Stem-loop Accession # MI0002801
          Stem-loop Sequence
          GGCCAGCUGUGAGUGUUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGCAAUAGUAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUACACAUUGUGGGGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026586
          miRBase ID: mml-miR-34a-3p
          Mature miRNA Sequence: AAUCAGCAAGUAUACUGCCCUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464787_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19468
          Ambion® Pre-miR™ miRNA Precursor : PM19468
          mirVana® miRNA inhibitor : MH19468
          mirVana® miRNA mimic : MC19468
          TaqMan™ Advanced miRNA Assay : rno481369_mir


          miRBase Alias:

          Rat: rno-miR-34a* (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline