Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM10381
          Assay Name: hsa-miR-181a-3p
          miRBase Accession Number: MI0000289
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 198859044 - 198859153 [-] on Build GRCh38
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Species: Human, Mouse, Rat, Alligator mississippiensis, Chicken, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Fugu rubripes, Gorilla gorilla, Guinea pig, Lagothrix lagotricha, Macaca nemestrina, Ornithorhynchus anatinus, Pan paniscus, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Taeniopygia guttata, Tetraodon nigroviridis, Zebrafish
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM10381
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000007, mir-181

          Mature miRNA Sequence:

          ACCAUCGACCGUUGAUUGUACC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-181a-3p MIMAT0000270
          Mouse mmu-miR-181a-1-3p MIMAT0000660
          Rat rno-miR-181a-1-3p MIMAT0000884
          Alligator mississippiensis ami-miR-181a-3p MIMAT0038187
          Chicken gga-miR-181a-3p MIMAT0001150
          Columba livia cli-miR-181a-3p MIMAT0038551
          View More Rows
          Cricetulus griseus cgr-miR-181a-3p MIMAT0023798
          Dasypus novemcinctus dno-miR-181a-3p MIMAT0047690
          Fugu rubripes fru-miR-181a-3p MIMAT0003005
          Gorilla gorilla ggo-miR-181a-3p MIMAT0002629
          Guinea pig cpo-miR-181a-3p MIMAT0047041
          Lagothrix lagotricha lla-miR-181a-3p MIMAT0002632
          Macaca nemestrina mne-miR-181a-3p MIMAT0002634
          Ornithorhynchus anatinus oan-miR-181a-1-3p MIMAT0006834
          Pan paniscus ppa-miR-181a-3p MIMAT0002636
          Pan troglodytes ptr-miR-181a-3p MIMAT0002624
          Pongo pygmaeus ppy-miR-181a-3p MIMAT0002627
          Pteropus alecto pal-miR-181a-3p MIMAT0039817
          Rabbit ocu-miR-181a-3p MIMAT0048276
          Rhesus monkey mml-miR-181a-3p MIMAT0002622
          Taeniopygia guttata tgu-miR-181a-1-3p MIMAT0014641
          Tetraodon nigroviridis tni-miR-181a-3p MIMAT0003006
          Zebrafish dre-miR-181a-3p MIMAT0001282

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-181a-1
          Stem-loop Accession # MI0000289
          Stem-loop Sequence
          UGAGUUUUGAGGUUGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUAAAAUCAAAACCAUCGACCGUUGAUUGUACCCUAUGGCUAACCAUCAUCUACUCCA
          Chromosome Location Chr. 1 - 198859044 - 198859153 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000270
          miRBase ID: hsa-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: Chr. 1 - 198859044 - 198859153 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-181a-1
          Stem-loop Accession # MI0000697
          Stem-loop Sequence
          GGUUGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUCAAAUAAAAACCAUCGACCGUUGAUUGUACCCUAUAGCUAACC
          Chromosome Location Chr. 1 - 137966455 - 137966541 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000660
          miRBase ID: mmu-miR-181a-1-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: Chr. 1 - 137966455 - 137966541 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-181a-1
          Stem-loop Accession # MI0000953
          Stem-loop Sequence
          AGGUUGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUCAAAUAAAAACCAUCGACCGUUGAUUGUACCCUAUAGCUAACCAUUAUCUACUCC
          Chromosome Location Chr. 13 - 54952742 - 54952841 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000884
          miRBase ID: rno-miR-181a-1-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: Chr. 13 - 54952742 - 54952841 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-181a-1
          Stem-loop Accession # MI0029761
          Stem-loop Sequence
          GUGGUUGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAUUUAAAGUGAAAACCAUCGACCGUUGAUUGUACCCUCCAGCUAACCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038187
          miRBase ID: ami-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-181a-1
          Stem-loop Accession # MI0001218
          Stem-loop Sequence
          UGUAGUGGUUGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUUAAGUGAAAACCAUCGACCGUUGAUUGUACCCUCCAGCUAACCAUCCUCCUCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001150
          miRBase ID: gga-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-181a-1
          Stem-loop Accession # MI0029989
          Stem-loop Sequence
          UGUAGUGGUUGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUAAAGUGAAAACCAUCGACCGUUGAUUGUACCCUCCAGCUAACCAUCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038551
          miRBase ID: cli-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-181a
          Stem-loop Accession # MI0020420
          Stem-loop Sequence
          GCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUCAAAUAAAAACCAUCGACCGUUGAUUGUACCCUAUAGCUAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023798
          miRBase ID: cgr-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-181a-1
          Stem-loop Accession # MI0039007
          Stem-loop Sequence
          AACAUUCAACGCUGUCGGUGAGUUUGGAAUUAAAAUCAAAACCAUCGACCGUUGAUUGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047690
          miRBase ID: dno-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-181a-1
          Stem-loop Accession # MI0003333
          Stem-loop Sequence
          GCUCGCCCCGGUGAACAUUCAACGCUGUCGGUGAGUUUGAGCUAAAUUGGAAAAACCAUCGACCGUUGAUUGUACCCCGUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003005
          miRBase ID: fru-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-181a-1
          Stem-loop Accession # MI0002937
          Stem-loop Sequence
          UGAGUUUUGAGGUUGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUAAAAUCAAAACCAUCGACCGUUGAUUGUACCCUAUGGCUAACCAUCAUCUUCUCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002629
          miRBase ID: ggo-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-181a-1
          Stem-loop Accession # MI0038665
          Stem-loop Sequence
          AACAUUCAACGCUGUCGGUGAGUUUGGAAUUAAAAUCAAAACCAUCGACCGUUGAUUGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047041
          miRBase ID: cpo-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Lagothrix lagotricha

          Stem-loop ID lla-mir-181a-1
          Stem-loop Accession # MI0002939
          Stem-loop Sequence
          UGAGUUUUGAGGUUGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUAAAAUCAAAACCAUCGACCGUUGAUUGUACCCUAUGGCUAACCAUCAUCUACUCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002632
          miRBase ID: lla-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-181a-1
          Stem-loop Accession # MI0002941
          Stem-loop Sequence
          UGAGUUUUGAGGUUGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUAAAAUCAAAACCAUCGACCGUUGAUUGUACCCUAUGGCUAACCAUCAUCUACUCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002634
          miRBase ID: mne-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-181a-1
          Stem-loop Accession # MI0006685
          Stem-loop Sequence
          GAAUUCUGCUUCGAAUGGUUGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUAAAGUAGAAACCAUCGACCGUUGAUUGUACCCUGCGGCAACCACCGUCCUCCUUUCUUCAUGCUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006834
          miRBase ID: oan-miR-181a-1-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-181a-1
          Stem-loop Accession # MI0002943
          Stem-loop Sequence
          UGAGUUUUGAGGUUGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUAAAAUCAAAACCAUCGACCGUUGAUUGUACCCUAUGGCUAACCAUCAUCUACUCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002636
          miRBase ID: ppa-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-181a-1
          Stem-loop Accession # MI0002933
          Stem-loop Sequence
          UGAGUUUUGAGGUUGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUAAAAUCAAAACCAUCGACCGUUGAUUGUACCCUAUGGCUAACCAUCAUCUACUCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002624
          miRBase ID: ptr-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-181a-1
          Stem-loop Accession # MI0002935
          Stem-loop Sequence
          UGAGUUUUGAGGUUGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUAAAAUCAAAACCAUCGACCGUUGAUUGUACCCUAUGGCUAACCAUCAUCUACUCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002627
          miRBase ID: ppy-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-181a-1
          Stem-loop Accession # MI0032344
          Stem-loop Sequence
          AACAUUCAACGCUGUCGGUGAGUUUGGAAUUAAAGUCAAAACCAUCGACCGUUGAUUGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039817
          miRBase ID: pal-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-181a-1
          Stem-loop Accession # MI0039315
          Stem-loop Sequence
          AACAUUCAACGCUGUCGGUGAGUUUGGAAUUAAAAUCAAAACCAUCGACCGUUGAUUGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048276
          miRBase ID: ocu-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-181a-1
          Stem-loop Accession # MI0002931
          Stem-loop Sequence
          UGAGUUUUGAGGUUGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUAAAAUCAAAACCAUCGACCGUUGAUUGUACCCUAUGGCUAACCAUCAUCUACUCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002622
          miRBase ID: mml-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-181a-1
          Stem-loop Accession # MI0013760
          Stem-loop Sequence
          UGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUAAAGUGAAAACCAUCGACCGUUGAUUGUACCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014641
          miRBase ID: tgu-miR-181a-1-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-181a-1
          Stem-loop Accession # MI0003334
          Stem-loop Sequence
          GCUCGCCCCGGUGAACAUUCAACGCUGUCGGUGAGUUUGAGCCAAAUUGGAAAAACCAUCGACCGUUGAUUGUACCCCGUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003006
          miRBase ID: tni-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-181a-1
          Stem-loop Accession # MI0001380
          Stem-loop Sequence
          GUUUGCCUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGAGCUAAAUGGAAAAAAACCAUCGACCGUUGAUUGUACCCUGCGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001282
          miRBase ID: dre-miR-181a-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307120_pri
          TaqMan™ Pri-miRNA Assay : Hs03302966_pri
          TaqMan™ Pri-miRNA Assay : Rn03465114_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10381
          TaqMan™ MicroRNA Assay : 000516
          mirVana® miRNA inhibitor : MH10381
          mirVana® miRNA mimic : MC10381
          TaqMan™ Advanced miRNA Assay : 479405_mir


          miRBase Alias:

          Human: hsa-miR-181a* (v17)
          Human: hsa-miR-213 (v8)
          Mouse: mmu-miR-213 (v8)
          Mouse: mmu-miR-181a-1* (v17)
          Mouse: mmu-miR-181a* (v9.2)
          Rat: rno-miR-213 (v8)
          Rat: rno-miR-181a-1* (v18)
          Rat: rno-miR-181a* (v15)
          Danio rerio: dre-miR-181a* (v18)
          Fugu rubripes: fru-miR-181a* (v18)
          Gallus gallus: gga-miR-213 (v8)
          Gallus gallus: gga-miR-181a* (v18)
          Gorilla gorilla adenovirus B7: ggo-miR-213 (v8)
          Gorilla gorilla adenovirus B7: ggo-miR-181a* (v18)
          Lactococcus lactis subsp. tructae: lla-miR-181a* (v18)
          Lactococcus lactis subsp. tructae: lla-miR-213 (v8)
          Macaca mulatta: mml-miR-213 (v8)
          Macaca mulatta: mml-miR-181a* (v18)
          Macaca nemestrina: mne-miR-213 (v8)
          Macaca nemestrina: mne-miR-181a* (v18)
          Ochrobactrum anthropi: oan-miR-181a-3p (v19)
          Ochrobactrum anthropi: oan-miR-181a* (v18)
          Pan troglodytes adenovirus: ptr-miR-213 (v8)
          Pan troglodytes adenovirus: ptr-miR-181a* (v18)
          Pectobacterium parvum: ppa-miR-181a* (v18)
          Pectobacterium parvum: ppa-miR-213 (v8)
          Pongo pygmaeus: ppy-miR-213 (v8)
          Pongo pygmaeus: ppy-miR-181a* (v18)
          Tetraodon nigroviridis: tni-miR-181a* (v18)
          Tetraodon nigroviridis: tni-miR-213 (v8)
          Trabulsiella guamensis ATCC 49490: tgu-miR-181a* (v18)
          Trabulsiella guamensis ATCC 49490: tgu-miR-181a-3p (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lq2bv:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0