Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03307120_pri
          Assay Name:
          mmu-mir-181a-1
          Stem-loop ID:
          mmu-mir-181a-1 mmu-mir-181b-1
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.1: 137966455 - 137966541 [+] on Build GRCm38
          Stem-loop Location:
          Chr.1: 137966455 - 137966541 [+] on Build GRCm38 Chr.1: 137966639 - 137966718 [+] on Build GRCm38
          Mature miRNA ID:
          mmu-miR-181a-1-3p mmu-miR-181a-5p mmu-miR-181b-5p mmu-miR-181b-1-3p
          Species:
          Mouse
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Mm03307120_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          88

          Chromosome Location:

          Chr. 1 - 137966256 - 137966256 [-] on 137966256 Build GRCm38

          Species:

          Mouse

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-181a-1-3p MIMAT0000660
          Mouse mmu-miR-181a-5p MIMAT0000210
          Mouse mmu-miR-181b-5p MIMAT0000673
          Mouse mmu-miR-181b-1-3p MIMAT0017067

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-181a-1
          Stem-loop Accession # MI0000697
          Stem-loop Sequence
          GGUUGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUCAAAUAAAAACCAUCGACCGUUGAUUGUACCCUAUAGCUAACC
          Chromosome Location Chr. 1 - 137966455 - 137966541 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000660
          miRBase ID: mmu-miR-181a-1-3p
          Mature miRNA Sequence: ACCAUCGACCGUUGAUUGUACC
          Chromosome Location: Chr. 1 - 137966455 - 137966541 [+] on Build GRCm38

          miRBase Accession #: MIMAT0000210
          miRBase ID: mmu-miR-181a-5p
          Mature miRNA Sequence: AACAUUCAACGCUGUCGGUGAGU
          Chromosome Location: Chr. 1 - 137966455 - 137966541 [+] on Build GRCm38

          Gene Family ID MIPF0000007, mir-181
          Distance to Amplicon 157 bases
          Stem-loop ID mmu-mir-181b-1
          Stem-loop Accession # MI0000723
          Stem-loop Sequence
          AGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUAGAAAAGCUCACUGAACAAUGAAUGCAACUGUGGCC
          Chromosome Location Chr. 1 - 137966639 - 137966718 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000673
          miRBase ID: mmu-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: Chr. 1 - 137966639 - 137966718 [+] on Build GRCm38

          miRBase Accession #: MIMAT0017067
          miRBase ID: mmu-miR-181b-1-3p
          Mature miRNA Sequence: CUCACUGAACAAUGAAUGCAA
          Chromosome Location: Chr. 1 - 137966639 - 137966718 [+] on Build GRCm38

          Gene Family ID MIPF0000007, mir-181
          Distance to Amplicon 341 bases

          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 245498_mat
          mirVana® miRNA inhibitor : MH10764
          mirVana® miRNA mimic : MC10764
          Ambion® Anti-miR™ miRNA Inhibitor : AM10381
          Ambion® Pre-miR™ miRNA Precursor : PM10381
          TaqMan™ MicroRNA Assay : 000516
          mirVana® miRNA inhibitor : MH10381
          mirVana® miRNA mimic : MC10381
          TaqMan™ Advanced miRNA Assay : 479405_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10421
          Ambion® Pre-miR™ miRNA Precursor : PM10421
          TaqMan™ MicroRNA Assay : 000480
          mirVana® miRNA inhibitor : MH10421
          mirVana® miRNA mimic : MC10421
          TaqMan™ Advanced miRNA Assay : 477857_mir
          TaqMan™ Advanced miRNA Assay : rno481485_mir
          TaqMan™ Advanced miRNA Assay : mmu481485_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM20327
          Ambion® Pre-miR™ miRNA Precursor : PM20327
          TaqMan™ MicroRNA Assay : 462578_mat
          mirVana® miRNA inhibitor : MH20327
          mirVana® miRNA mimic : MC20327


          miRBase Alias:

          Mouse: mmu-mir-181b-1 (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0