Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10764
          Assay Name: mmu-miR-181b-5p
          miRBase Accession Number: MI0000723
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 137966639 - 137966718 [+] on Build GRCm38
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Species: Mouse, Alligator mississippiensis, Bovine, Chrysemys picta, Columba livia, Dasypus novemcinctus, Gadus morhua, Gorilla gorilla, Guinea pig, Hippoglossus hippoglossus, Lagothrix lagotricha, Macaca nemestrina, Oryzias latipes, Pan paniscus, Pig, Pteropus alecto, Rabbit, Rhesus monkey, Taeniopygia guttata
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10764

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000007, mir-181

          Mature miRNA Sequence:

          AACAUUCAUUGCUGUCGGUGGGUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-181b-5p MIMAT0000673
          Alligator mississippiensis ami-miR-181b-5p MIMAT0038190
          Bovine bta-miR-181b MIMAT0003793
          Chrysemys picta cpi-miR-181b-5p MIMAT0037795
          Columba livia cli-miR-181b-5p MIMAT0038553
          Dasypus novemcinctus dno-miR-181b-5p MIMAT0047692
          View More Rows
          Gadus morhua gmo-miR-181b-5p MIMAT0044134
          Gorilla gorilla ggo-miR-181b MIMAT0002630
          Guinea pig cpo-miR-181b-5p MIMAT0047043
          Hippoglossus hippoglossus hhi-miR-181b MIMAT0030353
          Lagothrix lagotricha lla-miR-181b MIMAT0002633
          Macaca nemestrina mne-miR-181b MIMAT0002635
          Oryzias latipes ola-miR-181b-5p MIMAT0022540
          Pan paniscus ppa-miR-181b MIMAT0002637
          Pig ssc-miR-181b MIMAT0002126
          Pteropus alecto pal-miR-181b-5p MIMAT0039844
          Rabbit ocu-miR-181b-5p MIMAT0048278
          Rhesus monkey mml-miR-181b-5p MIMAT0002623
          Taeniopygia guttata tgu-miR-181b-5p MIMAT0014563

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-181b-1
          Stem-loop Accession # MI0000723
          Stem-loop Sequence
          AGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUAGAAAAGCUCACUGAACAAUGAAUGCAACUGUGGCC
          Chromosome Location Chr. 1 - 137966639 - 137966718 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000673
          miRBase ID: mmu-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: Chr. 1 - 137966639 - 137966718 [+] on Build GRCm38

          Stem-loop ID mmu-mir-181b-2
          Stem-loop Accession # MI0000823
          Stem-loop Sequence
          UUGAUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUGAAUGUCAACCAACUCACUGAUCAAUGAAUGCAAACUGCGGGCCAAAAA
          Chromosome Location Chr. 2 - 38853830 - 38853918 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000673
          miRBase ID: mmu-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: Chr. 1 - 137966639 - 137966718 [+] on Build GRCm38

          Alligator mississippiensis

          Stem-loop ID ami-mir-181b-1
          Stem-loop Accession # MI0029764
          Stem-loop Sequence
          AAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUUAACUAUGCGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038190
          miRBase ID: ami-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Stem-loop ID ami-mir-181b-2
          Stem-loop Accession # MI0029765
          Stem-loop Sequence
          UGCAUCUAAUGGCUGCAAUCAACAUUCAUUGCUGUCGGUGGGUUUUAGUUUGUACCAACUCACUGAUCAAUGAAUGCAAACUGCGGACCAGAUAGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038190
          miRBase ID: ami-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Stem-loop ID ami-mir-181b-3
          Stem-loop Accession # MI0029766
          Stem-loop Sequence
          AAAAAGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGUGAUGCUGAAGGGAAAACUCACGGAUCAAUGAAUGCAACUGUGAUUGGAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038190
          miRBase ID: ami-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-181b-1
          Stem-loop Accession # MI0010485
          Stem-loop Sequence
          CUUGGGCAGAGGUUCUUUCUUAAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAGUGCAACUGUGGCCCCGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003793
          miRBase ID: bta-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Stem-loop ID bta-mir-181b-2
          Stem-loop Accession # MI0005013
          Stem-loop Sequence
          CUGAUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUGACUUUGAAUCAACUCACUGAUCAAUGAAUGUAAACUGCGGACCAAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003793
          miRBase ID: bta-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-181b-1
          Stem-loop Accession # MI0029503
          Stem-loop Sequence
          AGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUUAACUAUGUGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037795
          miRBase ID: cpi-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Stem-loop ID cpi-mir-181b-2
          Stem-loop Accession # MI0029504
          Stem-loop Sequence
          GCAUCUAAUGGCUGCAAUCAACAUUCAUUGCUGUCGGUGGGUUUUAGUUUGUAUCAACUCACUGAUCAAUGAAUGCAAACUGCGGACCAGGUAGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037795
          miRBase ID: cpi-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Stem-loop ID cpi-mir-181b-3
          Stem-loop Accession # MI0029505
          Stem-loop Sequence
          CGAAAAGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGUGUUGUUGAAGGAAAAACUCACUGAUCAAUGAAUGCAACUGUGAUUGGAAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037795
          miRBase ID: cpi-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-181b-1
          Stem-loop Accession # MI0029991
          Stem-loop Sequence
          AAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUUAACUAUGUGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038553
          miRBase ID: cli-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Stem-loop ID cli-mir-181b-2
          Stem-loop Accession # MI0029992
          Stem-loop Sequence
          CUAAUGGCUGCAAUCAACAUUCAUUGCUGUCGGUGGGUUUUCUUUUCUGUCAACUCACUGAUCAAUGAAUGCAAACUGCGGACCAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038553
          miRBase ID: cli-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-181b-1
          Stem-loop Accession # MI0039009
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAAUGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047692
          miRBase ID: dno-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Stem-loop ID dno-mir-181b-2
          Stem-loop Accession # MI0039010
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUUGCAUCCGAAUCAACUCGCUGAUCAAUGAAUGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047692
          miRBase ID: dno-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-181b-1
          Stem-loop Accession # MI0036066
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUUGGCUAUGAAGAAAAGCUCACUGAACGAUGAAUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044134
          miRBase ID: gmo-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Stem-loop ID gmo-mir-181b-2
          Stem-loop Accession # MI0036211
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUUCUGGUUGAAAUGACUCAUUGACGAAUGAAUGCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044134
          miRBase ID: gmo-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-181b
          Stem-loop Accession # MI0002938
          Stem-loop Sequence
          CCUGUGCAGAGAUUAUUUUUUAAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCGCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002630
          miRBase ID: ggo-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-181b-1
          Stem-loop Accession # MI0038667
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUGAAUUGUGUGGACAAGCUCACUGGACAAUGAAUGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047043
          miRBase ID: cpo-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Stem-loop ID cpo-mir-181b-2
          Stem-loop Accession # MI0038668
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUUGAAUCUGAAUUGACUCACUGAUCAAUGAAUGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047043
          miRBase ID: cpo-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Hippoglossus hippoglossus

          Stem-loop ID hhi-mir-181b
          Stem-loop Accession # MI0025431
          Stem-loop Sequence
          AGGAGGCGUUCACUCUCAAAGGCUGCAAUAAACAUUCAUUGCUGUCGGUGGGUUUACAUAAGAAUAACUCACUGAUCAAUGAAUGCAGACUGCGGUUCAAACACCUCAGUAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0030353
          miRBase ID: hhi-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Lagothrix lagotricha

          Stem-loop ID lla-mir-181b
          Stem-loop Accession # MI0002940
          Stem-loop Sequence
          CCUGUGCAGAGAUUAUUUUUUAAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCGCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002633
          miRBase ID: lla-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-181b
          Stem-loop Accession # MI0002942
          Stem-loop Sequence
          CCUGUGCAGAGAUUAUUUUUUAAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUAGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCGCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002635
          miRBase ID: mne-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Oryzias latipes

          Stem-loop ID ola-mir-181b-1
          Stem-loop Accession # MI0019423
          Stem-loop Sequence
          AGCAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUUAACUAUGUGGAAGAGCUCACUGAACGAUGAAUGCAACUGUGUCCCAGAUUUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022540
          miRBase ID: ola-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Stem-loop ID ola-mir-181b-2
          Stem-loop Accession # MI0019466
          Stem-loop Sequence
          UCCAGGAGUCAACCUUAAGGCUGCAAUAAACAUUCAUUGCUGUCGGUGGGUUUAUGCAGGAACAACUCACUGAUCAAUGAAUGCAAACUGCGGUUCGCACAUCCUGCUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022540
          miRBase ID: ola-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-181b
          Stem-loop Accession # MI0002944
          Stem-loop Sequence
          CCUGUGCAGAGAUUAUUUUUUAAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCGCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002637
          miRBase ID: ppa-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-181b-1
          Stem-loop Accession # MI0013107
          Stem-loop Sequence
          GGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUAGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002126
          miRBase ID: ssc-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Stem-loop ID ssc-mir-181b-2
          Stem-loop Accession # MI0002420
          Stem-loop Sequence
          AUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUGAGUCUGAAUCAACUCACUGAUCAAUGAAUGCAAACUGCGGACCAAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002126
          miRBase ID: ssc-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-181b-1
          Stem-loop Accession # MI0032363
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGAUGAGCUCACUGAACAAUGAAUGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039844
          miRBase ID: pal-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Stem-loop ID pal-mir-181b-2
          Stem-loop Accession # MI0032358
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUUGAGUCUGAAUCAACUCACUGAUCAAUGAAUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039844
          miRBase ID: pal-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-181b-1
          Stem-loop Accession # MI0039317
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAAUGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048278
          miRBase ID: ocu-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Stem-loop ID ocu-mir-181b-2
          Stem-loop Accession # MI0039318
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUUGAGUGCAAAUCAACUCACUGAUCAAUGAAUGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048278
          miRBase ID: ocu-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-181b-1
          Stem-loop Accession # MI0002932
          Stem-loop Sequence
          CCUGUGCAGAGAUUAUUUUUUAAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUAGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCGCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002623
          miRBase ID: mml-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Stem-loop ID mml-mir-181b-2
          Stem-loop Accession # MI0007646
          Stem-loop Sequence
          CUGAUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUGAGUCUGAAUCAACUCACUGAUCGAUGAAUGCAAACUGCGGACCAAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002623
          miRBase ID: mml-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-181b-1
          Stem-loop Accession # MI0013782
          Stem-loop Sequence
          GGUCACAAUGAACAUUCAUUGCUGUCGGUGGGUUUAACUAUGUGGACAAGCUCACUGAACAAUGAAUGCAACUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014563
          miRBase ID: tgu-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA

          Stem-loop ID tgu-mir-181b-2
          Stem-loop Accession # MI0013783
          Stem-loop Sequence
          GGCUGCAAUCAACAUUCAUUGCUGUCGGUGGGUUUUCAUUUCUAUCAACUCACUGACCAAUGAAUGCAAACUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014563
          miRBase ID: tgu-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307414_pri
          TaqMan™ Pri-miRNA Assay : Mm03307120_pri
          TaqMan™ MicroRNA Assay : 245498_mat
          mirVana® miRNA mimic : MC10764


          miRBase Alias:

          Macaca mulatta: mml-miR-181b (v19)
          Trabulsiella guamensis ATCC 49490: tgu-miR-181b (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0