Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH20327
          Assay Name: hsa-miR-181b-3p
          miRBase Accession Number: MI0000270
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 198858873 - 198858982 [-] on Build GRCh38
          Mature miRNA Sequence: CUCACUGAACAAUGAAUGCAA
          Species: Human, Mouse, Rat, Alligator mississippiensis, Columba livia, Dasypus novemcinctus, Pteropus alecto, Rabbit, Zebrafish
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH20327

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000007, mir-181

          Mature miRNA Sequence:

          CUCACUGAACAAUGAAUGCAA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-181b-3p MIMAT0022692
          Mouse mmu-miR-181b-1-3p MIMAT0017067
          Rat rno-miR-181b-1-3p MIMAT0017139
          Alligator mississippiensis ami-miR-181b-3p MIMAT0038191
          Columba livia cli-miR-181b-3p MIMAT0038554
          Dasypus novemcinctus dno-miR-181b-3p MIMAT0047693
          View More Rows
          Pteropus alecto pal-miR-181b-3p MIMAT0039852
          Rabbit ocu-miR-181b-3p MIMAT0048279
          Zebrafish dre-miR-181b-3-3p MIMAT0048656

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-181b-1
          Stem-loop Accession # MI0000270
          Stem-loop Sequence
          CCUGUGCAGAGAUUAUUUUUUAAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCGCUU
          Chromosome Location Chr. 1 - 198858873 - 198858982 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0022692
          miRBase ID: hsa-miR-181b-3p
          Mature miRNA Sequence: CUCACUGAACAAUGAAUGCAA
          Chromosome Location: Chr. 1 - 198858873 - 198858982 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-181b-1
          Stem-loop Accession # MI0000723
          Stem-loop Sequence
          AGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUAGAAAAGCUCACUGAACAAUGAAUGCAACUGUGGCC
          Chromosome Location Chr. 1 - 137966639 - 137966718 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017067
          miRBase ID: mmu-miR-181b-1-3p
          Mature miRNA Sequence: CUCACUGAACAAUGAAUGCAA
          Chromosome Location: Chr. 1 - 137966639 - 137966718 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-181b-1
          Stem-loop Accession # MI0000926
          Stem-loop Sequence
          CCUGUGCAGAGAUGAUGUUUACAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUAGAAAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCGCUU
          Chromosome Location Chr. 13 - 54952903 - 54953012 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017139
          miRBase ID: rno-miR-181b-1-3p
          Mature miRNA Sequence: CUCACUGAACAAUGAAUGCAA
          Chromosome Location: Chr. 13 - 54952903 - 54953012 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-181b-1
          Stem-loop Accession # MI0029764
          Stem-loop Sequence
          AAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUUAACUAUGCGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038191
          miRBase ID: ami-miR-181b-3p
          Mature miRNA Sequence: CUCACUGAACAAUGAAUGCAA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-181b-1
          Stem-loop Accession # MI0029991
          Stem-loop Sequence
          AAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUUAACUAUGUGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038554
          miRBase ID: cli-miR-181b-3p
          Mature miRNA Sequence: CUCACUGAACAAUGAAUGCAA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-181b-1
          Stem-loop Accession # MI0039009
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAAUGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047693
          miRBase ID: dno-miR-181b-3p
          Mature miRNA Sequence: CUCACUGAACAAUGAAUGCAA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-181b-1
          Stem-loop Accession # MI0032363
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGAUGAGCUCACUGAACAAUGAAUGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039852
          miRBase ID: pal-miR-181b-3p
          Mature miRNA Sequence: CUCACUGAACAAUGAAUGCAA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-181b-1
          Stem-loop Accession # MI0039317
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAAUGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048279
          miRBase ID: ocu-miR-181b-3p
          Mature miRNA Sequence: CUCACUGAACAAUGAAUGCAA
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-181b-3
          Stem-loop Accession # MI0039514
          Stem-loop Sequence
          AAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGGAUUGUAAAAGAAAGCUCACUGAACAAUGAAUGCAACUGUGUCCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048656
          miRBase ID: dre-miR-181b-3-3p
          Mature miRNA Sequence: CUCACUGAACAAUGAAUGCAA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465090_pri
          TaqMan™ Pri-miRNA Assay : Hs03302963_pri
          TaqMan™ Pri-miRNA Assay : Mm03307120_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM20327
          Ambion® Pre-miR™ miRNA Precursor : PM20327
          TaqMan™ MicroRNA Assay : 462578_mat
          mirVana® miRNA mimic : MC20327


          miRBase Alias:

          Rat: rno-miR-181b-1* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mfv9s:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0