Search
Search

| Assay Name: | mmu-miR-466b-3p |
|---|---|
| miRBase Accession Number: | MI0005502 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AUACAUACACGCACACAUAAGA |
| Species: | Mouse |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000208, mir-466 |
Mature miRNA Sequence: | AUACAUACACGCACACAUAAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-466b-3p | MIMAT0004876 |
| Mouse | mmu-miR-466c-3p | MIMAT0004878 |
| Mouse | mmu-miR-466p-3p | MIMAT0014892 |