Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm04272635_pri
Assay Name:
mmu-mir-466p
Stem-loop ID:
mmu-mir-466p
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.2: 10506006 - 10506094 [+] on Build GRCm38
Stem-loop Location:
Chr.2: 10506006 - 10506094 [+] on Build GRCm38
Mature miRNA ID:
mmu-miR-466p-5p
mmu-miR-466p-3p
Species:
Mouse
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Mm04272635_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
91
Chromosome Location:
Chr. 2 - 10506344 - 10506344 [-] on 10506344 Build GRCm38
Species:
Mouse
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Mouse
mmu-miR-466p-5p
MIMAT0014891
Mouse
mmu-miR-466p-3p
MIMAT0014892
Stem-loop Details
Mouse
Stem-loop ID
mmu-mir-466p
Stem-loop Accession #
MI0014078
Stem-loop Sequence
UGUGUAUAUGUGUUUAUGUGUGUGUACAUGUACAUGUGUGAAUAUGAUGAUAUAUACAUACACGCACACAUAAGACACAUAUGAGCACA
Chromosome Location
Chr. 2 - 10506006 - 10506094 [+] on Build GRCm38
Mature MicroRNA
miRBase Accession #: MIMAT0014891
miRBase ID: mmu-miR-466p-5p
Mature miRNA Sequence: UAUGUGUGUGUACAUGUACAU
Chromosome Location: Chr. 2 - 10506006 - 10506094 [+] on Build GRCm38
miRBase Accession #: MIMAT0014892
miRBase ID: mmu-miR-466p-3p
Mature miRNA Sequence: AUACAUACACGCACACAUAAGA
Chromosome Location: Chr. 2 - 10506006 - 10506094 [+] on Build GRCm38
Gene Family ID
MIPF0000208,
mir-466
Distance to Amplicon
202 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM19973
Ambion
®
Pre-miR
™
miRNA Precursor :
PM19973
TaqMan
™
MicroRNA Assay :
462364_mat
mir
Vana
®
miRNA inhibitor :
MH19973
mir
Vana
®
miRNA mimic :
MC19973
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM19359
Ambion
®
Pre-miR
™
miRNA Precursor :
PM19359
TaqMan
™
MicroRNA Assay :
464896_mat
mir
Vana
®
miRNA inhibitor :
MH19359
mir
Vana
®
miRNA mimic :
MC19359
TaqMan
™
Advanced miRNA Assay :
mmu482642_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline