Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03308706_pri
          Assay Name:
          mmu-mir-467e
          Stem-loop ID:
          mmu-mir-466b-3 mmu-mir-467a-10 mmu-mir-467e mmu-mir-669a-11 mmu-mir-669a-12
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.2: 10503565 - 10503645 [+] on Build GRCm38
          Stem-loop Location:
          Chr.2: 10503565 - 10503645 [+] on Build GRCm38 Chr.2: 10503273 - 10503355 [+] on Build GRCm38 Chr.2: 10505721 - 10505807 [+] on Build GRCm38 Chr.2: 10501339 - 10501425 [+] on Build GRCm38 Chr.2: 10503791 - 10503877 [+] on Build GRCm38
          Mature miRNA ID:
          mmu-miR-466b-3p mmu-miR-466b-5p mmu-miR-467a-5p mmu-miR-467a-3p mmu-miR-467e-3p mmu-miR-467e-5p mmu-miR-669a-5p mmu-miR-669a-3p
          Species:
          Mouse
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Mm03308706_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          110

          Chromosome Location:

          Chr. 2 - 10505517 - 10505517 [+] on 10505517 Build GRCm38

          Species:

          Mouse

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-466b-3p MIMAT0004876
          Mouse mmu-miR-466b-5p MIMAT0004875
          Mouse mmu-miR-467a-5p MIMAT0003409
          Mouse mmu-miR-467a-3p MIMAT0002108
          Mouse mmu-miR-467e-3p MIMAT0005294
          Mouse mmu-miR-467e-5p MIMAT0005293
          View More Rows
          Mouse mmu-miR-669a-5p MIMAT0003477
          Mouse mmu-miR-669a-3p MIMAT0017243

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-466b-3
          Stem-loop Accession # MI0005504
          Stem-loop Sequence
          UAUAUGUGUUGAUGUGUGUGUACAUGUACAUGUGUGAAUAUGAUAUACAAAUACAUACACGCACACAUAAGACACAUAUGA
          Chromosome Location Chr. 2 - 10503565 - 10503645 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004876
          miRBase ID: mmu-miR-466b-3p
          Mature miRNA Sequence: AUACAUACACGCACACAUAAGA
          Chromosome Location: Chr. 2 - 10503565 - 10503645 [+] on Build GRCm38

          miRBase Accession #: MIMAT0004875
          miRBase ID: mmu-miR-466b-5p
          Mature miRNA Sequence: UGAUGUGUGUGUACAUGUACAU
          Chromosome Location: Chr. 2 - 10503565 - 10503645 [+] on Build GRCm38

          Gene Family ID MIPF0000208, mir-466
          Distance to Amplicon 1800 bases
          Stem-loop ID mmu-mir-467a-10
          Stem-loop Accession # MI0014076
          Stem-loop Sequence
          UGUGCCCGUGUGCGUAAGUGCCUGCAUGUAUAUGCGUGUAUAUUUUAUGCAUAUACAUACACACACCUACACACACAUGCACA
          Chromosome Location Chr. 2 - 10503273 - 10503355 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003409
          miRBase ID: mmu-miR-467a-5p
          Mature miRNA Sequence: UAAGUGCCUGCAUGUAUAUGCG
          Chromosome Location: Chr. 2 - 10503273 - 10503355 [+] on Build GRCm38

          miRBase Accession #: MIMAT0002108
          miRBase ID: mmu-miR-467a-3p
          Mature miRNA Sequence: CAUAUACAUACACACACCUACA
          Chromosome Location: Chr. 2 - 10503273 - 10503355 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 2090 bases
          Stem-loop ID mmu-mir-467e
          Stem-loop Accession # MI0006128
          Stem-loop Sequence
          GCCUGUGUGCAUAAGUGUGAGCAUGUAUAUGUGUGUAUAUUUUAUGCAUAUACAUACACACACCUAUAUACACAUGCACACAGACAU
          Chromosome Location Chr. 2 - 10505721 - 10505807 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0005294
          miRBase ID: mmu-miR-467e-3p
          Mature miRNA Sequence: AUAUACAUACACACACCUAUAU
          Chromosome Location: Chr. 2 - 10505721 - 10505807 [+] on Build GRCm38

          miRBase Accession #: MIMAT0005293
          miRBase ID: mmu-miR-467e-5p
          Mature miRNA Sequence: AUAAGUGUGAGCAUGUAUAUGU
          Chromosome Location: Chr. 2 - 10505721 - 10505807 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 167 bases
          Stem-loop ID mmu-mir-669a-11
          Stem-loop Accession # MI0014075
          Stem-loop Sequence
          UGUGCAUGUGUGUAUAGUUGUGUGUGCAUGUUCAUGUCUAUAUUUGAAUAUACAUAACAUACACACACACGUAUAAACGCAAGCACA
          Chromosome Location Chr. 2 - 10501339 - 10501425 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003477
          miRBase ID: mmu-miR-669a-5p
          Mature miRNA Sequence: AGUUGUGUGUGCAUGUUCAUGUCU
          Chromosome Location: Chr. 2 - 10503791 - 10503877 [+] on Build GRCm38

          miRBase Accession #: MIMAT0017243
          miRBase ID: mmu-miR-669a-3p
          Mature miRNA Sequence: ACAUAACAUACACACACACGUAU
          Chromosome Location: Chr. 2 - 10503791 - 10503877 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 4020 bases
          Stem-loop ID mmu-mir-669a-12
          Stem-loop Accession # MI0014077
          Stem-loop Sequence
          UGUGCAUGUGUGUAUAGUUGUGUGUGCAUGUUCAUGUCUAUAUUUGAAUAUACAUAACAUACACACACACGUAUAAACGCAAGCACA
          Chromosome Location Chr. 2 - 10503791 - 10503877 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003477
          miRBase ID: mmu-miR-669a-5p
          Mature miRNA Sequence: AGUUGUGUGUGCAUGUUCAUGUCU
          Chromosome Location: Chr. 2 - 10503791 - 10503877 [+] on Build GRCm38

          miRBase Accession #: MIMAT0017243
          miRBase ID: mmu-miR-669a-3p
          Mature miRNA Sequence: ACAUAACAUACACACACACGUAU
          Chromosome Location: Chr. 2 - 10503791 - 10503877 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 1568 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM12667
          Ambion® Pre-miR™ miRNA Precursor : PM12667
          TaqMan™ MicroRNA Assay : 002569
          mirVana® miRNA inhibitor : MH12667
          mirVana® miRNA mimic : MC12667
          Ambion® Anti-miR™ miRNA Inhibitor : AM12611
          Ambion® Pre-miR™ miRNA Precursor : PM12611
          TaqMan™ MicroRNA Assay : 002568
          mirVana® miRNA inhibitor : MH12611
          mirVana® miRNA mimic : MC12611
          Ambion® Anti-miR™ miRNA Inhibitor : AM12639
          Ambion® Pre-miR™ miRNA Precursor : PM12639
          TaqMan™ MicroRNA Assay : 002587
          mirVana® miRNA inhibitor : MH12639
          mirVana® miRNA mimic : MC12639
          TaqMan™ Advanced miRNA Assay : mmu483234_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM20509
          Ambion® Pre-miR™ miRNA Precursor : PM20509
          mirVana® miRNA inhibitor : MH20509
          mirVana® miRNA mimic : MC20509
          TaqMan™ Advanced miRNA Assay : mmu482917_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM19687
          Ambion® Pre-miR™ miRNA Precursor : PM19687
          TaqMan™ MicroRNA Assay : 464179_mat
          mirVana® miRNA inhibitor : MH19687
          mirVana® miRNA mimic : MC19687
          TaqMan™ Advanced miRNA Assay : mmu482998_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM19954
          Ambion® Pre-miR™ miRNA Precursor : PM19954
          TaqMan™ MicroRNA Assay : 463964_mat
          mirVana® miRNA inhibitor : MH19954
          mirVana® miRNA mimic : MC19954
          Ambion® Anti-miR™ miRNA Inhibitor : AM19359
          Ambion® Pre-miR™ miRNA Precursor : PM19359
          TaqMan™ MicroRNA Assay : 464896_mat
          mirVana® miRNA inhibitor : MH19359
          mirVana® miRNA mimic : MC19359
          TaqMan™ Advanced miRNA Assay : mmu482642_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM19674
          Ambion® Pre-miR™ miRNA Precursor : PM19674
          TaqMan™ MicroRNA Assay : 462897_mat
          mirVana® miRNA inhibitor : MH19674
          mirVana® miRNA mimic : MC19674

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline