Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu481304_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-34a-5p
          Stem-loop Accession Number: MI0000268
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 9151668 - 9151777 [-] on Build GRCh38
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Species: Human, Mouse, Rat, Astatotilapia burtoni, Ateles geoffroyi, Bovine, Chrysemys picta, Columba livia, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Dog, Gadus morhua, Goat, Gorilla gorilla, Guinea pig, Horse, Ictalurus punctatus, Lagothrix lagotricha, Macaca nemestrina, Metriaclima zebra, Microcebus murinus, Neolamprologus brichardi, Nomascus leucogenys, Oreochromis niloticus, Otolemur garnettii, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Pundamilia nyererei, Python bivittatus, Rabbit, Rhesus monkey, Saguinus labiatus, Taeniopygia guttata, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu481304_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000039, mir-34

          Mature miRNA Sequence:

          UGGCAGUGUCUUAGCUGGUUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-34a-5p MIMAT0000255
          Mouse mmu-miR-34a-5p MIMAT0000542
          Rat rno-miR-34a-5p MIMAT0000815
          Astatotilapia burtoni abu-miR-34 MIMAT0042147
          Ateles geoffroyi age-miR-34a MIMAT0002495
          Bovine bta-miR-34a MIMAT0004340
          View More Rows
          Chrysemys picta cpi-miR-34a-5p MIMAT0037692
          Columba livia cli-miR-34a-5p MIMAT0038467
          Cricetulus griseus cgr-miR-34a MIMAT0023926
          Cyprinus carpio ccr-miR-34 MIMAT0026292
          Dasypus novemcinctus dno-miR-34a-5p MIMAT0047575
          Dog cfa-miR-34a MIMAT0006690
          Gadus morhua gmo-miR-34-5p MIMAT0044295
          Goat chi-miR-34a MIMAT0036166
          Gorilla gorilla ggo-miR-34a MIMAT0002494
          Guinea pig cpo-miR-34a-5p MIMAT0046922
          Horse eca-miR-34a MIMAT0012917
          Ictalurus punctatus ipu-miR-34a MIMAT0029503
          Lagothrix lagotricha lla-miR-34a MIMAT0002501
          Macaca nemestrina mne-miR-34a MIMAT0002502
          Metriaclima zebra mze-miR-34 MIMAT0042398
          Microcebus murinus mmr-miR-34a MIMAT0049178
          Neolamprologus brichardi nbr-miR-34 MIMAT0042579
          Nomascus leucogenys nle-miR-34a MIMAT0049383
          Oreochromis niloticus oni-miR-34 MIMAT0042745
          Otolemur garnettii oga-miR-34a MIMAT0049615
          Pan paniscus ppa-miR-34a MIMAT0002496
          Pan troglodytes ptr-miR-34a MIMAT0002498
          Pig ssc-miR-34a MIMAT0007757
          Pongo pygmaeus ppy-miR-34a MIMAT0002497
          Pteropus alecto pal-miR-34a-5p MIMAT0040037
          Pundamilia nyererei pny-miR-34 MIMAT0042962
          Python bivittatus pbv-miR-34a-5p MIMAT0038884
          Rabbit ocu-miR-34a-5p MIMAT0048167
          Rhesus monkey mml-miR-34a-5p MIMAT0002499
          Saguinus labiatus sla-miR-34a MIMAT0002500
          Taeniopygia guttata tgu-miR-34a MIMAT0014598
          Zebrafish dre-miR-34a MIMAT0001269

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-34a
          Stem-loop Accession # MI0000268
          Stem-loop Sequence
          GGCCAGCUGUGAGUGUUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGCAAUAGUAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUGCACGUUGUGGGGCCC
          Chromosome Location Chr. 1 - 9151668 - 9151777 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000255
          miRBase ID: hsa-miR-34a-5p
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: Chr. 1 - 9151668 - 9151777 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-34a
          Stem-loop Accession # MI0000584
          Stem-loop Sequence
          CCAGCUGUGAGUAAUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGUAUUAGCUAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUGCACAUUGU
          Chromosome Location Chr. 4 - 150068454 - 150068555 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000542
          miRBase ID: mmu-miR-34a-5p
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: Chr. 4 - 150068454 - 150068555 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-34a
          Stem-loop Accession # MI0000877
          Stem-loop Sequence
          CCGGCUGUGAGUAAUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGUAUUAGCUAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUGCACGUUGU
          Chromosome Location Chr. 5 - 167092491 - 167092592 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000815
          miRBase ID: rno-miR-34a-5p
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: Chr. 5 - 167092491 - 167092592 [+] on Build Rnor_6.0

          Astatotilapia burtoni

          Stem-loop ID abu-mir-34
          Stem-loop Accession # MI0033862
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGAGGAGUGAGAAUGAAGCAAUCAGCAAGUAUACUGCCGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042147
          miRBase ID: abu-miR-34
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Ateles geoffroyi

          Stem-loop ID age-mir-34a
          Stem-loop Accession # MI0002797
          Stem-loop Sequence
          GGCCAGCUGUGAGUGUUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGUGCAAUAGUGAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUGCACGUUGUGGGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002495
          miRBase ID: age-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-34a
          Stem-loop Accession # MI0005464
          Stem-loop Sequence
          GGCCAGCUGUGAGUGUUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGUAAUAAUGCAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUGCACGUUGUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004340
          miRBase ID: bta-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-34a
          Stem-loop Accession # MI0029434
          Stem-loop Sequence
          GUGAGUGUUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGCAAUAAUCAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUGCACAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037692
          miRBase ID: cpi-miR-34a-5p
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-34a
          Stem-loop Accession # MI0029939
          Stem-loop Sequence
          GUGUUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGCAAUAGUCAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038467
          miRBase ID: cli-miR-34a-5p
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-34a
          Stem-loop Accession # MI0020505
          Stem-loop Sequence
          AGUGCUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGUAAUAGCGAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023926
          miRBase ID: cgr-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-34
          Stem-loop Accession # MI0023392
          Stem-loop Sequence
          UCUUUGGCAGUGUCUUAGCUGGUUGUUGUGUGGAGUGAGAACGAAGCAAUCAGCAAGUAUACUGCCGCAGAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026292
          miRBase ID: ccr-miR-34
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-34a
          Stem-loop Accession # MI0038944
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGAGUAAUAGUGCAGGAAGCAAUCAGCAAGUAUACUGCCCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047575
          miRBase ID: dno-miR-34a-5p
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-34a
          Stem-loop Accession # MI0008101
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGAGUAAUAGUGAAGGAAGCAAUCAGCAAGUAUACUGCCCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006690
          miRBase ID: cfa-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-34
          Stem-loop Accession # MI0036191
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGAGGAGUGAGAACGAAGCAAUCAGCAAGUAUACUGCCGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044295
          miRBase ID: gmo-miR-34-5p
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-34a
          Stem-loop Accession # MI0030757
          Stem-loop Sequence
          GGCCAGCUGUGAGUGUUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGUAAUAAUGCAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUGCACGUUGUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036166
          miRBase ID: chi-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-34a
          Stem-loop Accession # MI0002796
          Stem-loop Sequence
          GGCCAGCUGUGAGUGUUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGCAAUAGUAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUGCACGUUGUGGGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002494
          miRBase ID: ggo-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-34a
          Stem-loop Accession # MI0038600
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGAGUAUUGGUGCAGGAAGCAAUCAGCAAGUAUACUGCCCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046922
          miRBase ID: cpo-miR-34a-5p
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-34a
          Stem-loop Accession # MI0012673
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGAGUAAUAGUGCGGGAAGCAAUCAGCAAGUAUACUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012917
          miRBase ID: eca-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-34a
          Stem-loop Accession # MI0024622
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGUGGAGUGAGAACGAAGCAAUCAGCAAGUAUACUGCCGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029503
          miRBase ID: ipu-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Lagothrix lagotricha

          Stem-loop ID lla-mir-34a
          Stem-loop Accession # MI0002803
          Stem-loop Sequence
          GGCCAGCUGUGAGUGUUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGCAAUAGUGAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUGCACGUUGUGGGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002501
          miRBase ID: lla-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-34a
          Stem-loop Accession # MI0002804
          Stem-loop Sequence
          GGCCAGCUGUGAGUGUUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGCAAUAGUAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUACACAUUGUGGGGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002502
          miRBase ID: mne-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Metriaclima zebra

          Stem-loop ID mze-mir-34
          Stem-loop Accession # MI0034190
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGAGGAGUGAGAAUGAAGCAAUCAGCAAGUAUACUGCCGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042398
          miRBase ID: mze-miR-34
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-34a
          Stem-loop Accession # MI0039896
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGAGAAAUGCAAAGGAAGCAAUCAGCAAGUAUACUGCCCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049178
          miRBase ID: mmr-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-34
          Stem-loop Accession # MI0034446
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGAGGAGUGAGAAUGAAGCAAUCAGCAAGUAUACUGCCGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042579
          miRBase ID: nbr-miR-34
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-34a
          Stem-loop Accession # MI0040121
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGAGCAAUAGUAAGGAAGCAAUCAGCAAGUAUACUGCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049383
          miRBase ID: nle-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-34
          Stem-loop Accession # MI0034671
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGAGGAGUGAGAACGAAGCAAUCAGCAAGUAUACUGCCGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042745
          miRBase ID: oni-miR-34
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-34a
          Stem-loop Accession # MI0040396
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGAGUAAUAGUGAAGGAAGCAAUCAGCAAGUAUACUGCCCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049615
          miRBase ID: oga-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-34a
          Stem-loop Accession # MI0002798
          Stem-loop Sequence
          GGCCAGCUGUGAGUGUUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGCAAUAGUAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUGCACGUUGUGGGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002496
          miRBase ID: ppa-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-34a
          Stem-loop Accession # MI0002800
          Stem-loop Sequence
          GGCCAGCUGUGAGUGUUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGCAAUAGUAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUGCACGUUGUGGGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002498
          miRBase ID: ptr-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-34a
          Stem-loop Accession # MI0008216
          Stem-loop Sequence
          UUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGUAAUAGCAAAGGAAGCAAUCAGCAGCAUCCUGCCCUAGAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007757
          miRBase ID: ssc-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-34a
          Stem-loop Accession # MI0002799
          Stem-loop Sequence
          GGCCAGCUGUGAGUGUUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGCAAUAGUAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUGCACGUUGUGGGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002497
          miRBase ID: ppy-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-34a
          Stem-loop Accession # MI0032461
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGAGCAAUGGUGAAGGAAGCAAUCAGCAAGUAUACUGCCCUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040037
          miRBase ID: pal-miR-34a-5p
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Pundamilia nyererei

          Stem-loop ID pny-mir-34
          Stem-loop Accession # MI0034952
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGAGGAGUGAGAAUGAAGCAAUCAGCAAGUAUACUGCCGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042962
          miRBase ID: pny-miR-34
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-34a
          Stem-loop Accession # MI0030193
          Stem-loop Sequence
          CUCUGUGAGUGCUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGUGCGAUGAUUGAAGCAGCAAUCAGCAAGCAUACUGCUGCAGAAGUACUGCACGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038884
          miRBase ID: pbv-miR-34a-5p
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-34a
          Stem-loop Accession # MI0039256
          Stem-loop Sequence
          UGGCAGUGUCUUAGCUGGUUGUUGUGAGUCACAGAGAAGGAAGCAAUCAGCAAGUAUACUGCCCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048167
          miRBase ID: ocu-miR-34a-5p
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-34a
          Stem-loop Accession # MI0002801
          Stem-loop Sequence
          GGCCAGCUGUGAGUGUUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGCAAUAGUAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUACACAUUGUGGGGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002499
          miRBase ID: mml-miR-34a-5p
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Saguinus labiatus

          Stem-loop ID sla-mir-34a
          Stem-loop Accession # MI0002802
          Stem-loop Sequence
          GGCCGGCUGUGAGUGUUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGCAAUAGUGAAGGAAGCAAUCAGCAAGUAUACUGCCCUAGAAGUGCUGCACGUUGUGGGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002500
          miRBase ID: sla-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-34a
          Stem-loop Accession # MI0013826
          Stem-loop Sequence
          AGUGUUUCUUUGGCAGUGUCUUAGCUGGUUGUUGUGAGCAAUAGUCAAGGAAGCAAUCAGCAAGUAUACUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014598
          miRBase ID: tgu-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-34a
          Stem-loop Accession # MI0001365
          Stem-loop Sequence
          CUGCUGUGAGUGGUUCUCUGGCAGUGUCUUAGCUGGUUGUUGUGUGGAGUGAGAACGAAGCAAUCAGCAAGUAUACUGCCGCAGAAACUCGUCACCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001269
          miRBase ID: dre-miR-34a
          Mature miRNA Sequence: UGGCAGUGUCUUAGCUGGUUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306897_pri
          TaqMan™ Pri-miRNA Assay : Rn03464787_pri
          TaqMan™ Pri-miRNA Assay : Hs03302884_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11030
          Ambion® Pre-miR™ miRNA Precursor : PM11030
          TaqMan™ MicroRNA Assay : 000426
          mirVana® miRNA inhibitor : MH11030
          mirVana® miRNA mimic : MC11030
          TaqMan™ Advanced miRNA Assay : rno481304_mir
          TaqMan™ Advanced miRNA Assay : 478048_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline