Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • FOXP3 Antibodies

          Invitrogen

          FOXP3 Monoclonal Antibody (236A/E7), eBioscience™

          Advanced Verification
          55 Published Figures
          167 References
          View all (120) FOXP3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite FOXP3 Monoclonal Antibody (236A/E7), eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          • Published Figures (55)
          • Advanced Verification (1)
          FOXP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FOXP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 60

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FOXP3 Antibody (14-4777-82) in IHC (P)

          Immunohistochemical analysis of FOXP3 was performed using formalin-fixed paraffin-embedded human lymph node [Follicular Grade: I (small cell)] tissue sections. To expose the target protein, heat-induced epitope retrieval was performed on de-paraffinized sections using eBioscience™ IHC Antigen Retrieval Solution - High pH (10X) (Product # 00-4956-58) diluted to 1X solution in water in a decloaking chamber at... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FOXP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FOXP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FOXP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FOXP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FOXP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FOXP3 Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
          FOXP3 Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
          FOXP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FOXP3 Antibody in Immunohistochemistry, Immunohistochemistry (Frozen) (IHC, IHC (F))
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          FOXP3 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry, Radioimmune assays (Flow, RIA)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody

          Product Details

          14-4777-82

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-10 µg/mL
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 79 publications 79 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1-5 µg/mL
          View 19 publications 19 publications

          Immunohistochemistry (Frozen) (IHC (F))

          5 µg/mL
          View 3 publications 3 publications

          Immunocytochemistry (ICC/IF)

          -
          View 6 publications 6 publications

          Flow Cytometry (Flow)

          -
          View 54 publications 54 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          Radioimmune Assays (RIA)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Non-human primate, Rhesus monkey

          Published species

          Cynomolgus monkey, Human, Mouse, Non-human primate, Rat, Rhesus monkey

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          236A/E7

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor Plus 647 
          • APC 
          • Biotin 
          • eFluor 450
          • eFluor 570
          • eFluor 660
          • FITC
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_467556

          Product Specific Information

          Description: The 236A/E7 antibody reacts with human Foxp3 protein also known as FORKHEAD BOX P3, SCURFIN, and JM2. Foxp3, a 49-55 kDa protein, is a member of the forkhead/winged-helix family of transcriptional regulators, and was identified as the gene defective in 'scurfy' (sf) mice. Constitutive high expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg cells), and ectopic expression of Foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

          Intracellular staining and flow cytometric analysis of freshly isolated human peripheral blood mononuclear cells (PBMCs) with the 236A/E7 antibody using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523) and protocol reveals staining of the CD4+CD25bright population.

          The epitope from 236A/E7 is different from that of PCH101 (Product # 72-5776-40). This antibody has also been shown to recognize rhesus macaque, sooty mangabey and cynomolgus macaque.

          Applications Tested: This 236A/E7 antibody has been tested by immunoblotting and immunohistochemistry of formalin-fixed paraffin embedded tissue using low or high pH antigen retrieval. For western blotting of reduced cell lysates (which include the nuclear fraction) the antibody can be used at 1-10 µg/mL and for IHC can be used at less than or equal to 5 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          FOXP3 (Forkhead box protein 3) is a member of the forkhead/winged-helix family of transcriptional regulators, highly conserved across mammals, and essential for normal immune homeostasis. FOXP3 is 381 amino acids long, stably and constitutively expressed at a high level in CD25 + CD4 positive regulatory T cells, a low level in CD4-positive/CD25-negative cells, and is absent in CD4-negative/CD8-positive T cells. FOXP3 may be a master regulatory gene, and a more specific marker of regulatory T cells. Defects in the gene encoding FOXP3 protein cause the scurfy phenotype in mice. In humans FOXP3 defects play a role in IPEX syndrome (immune dysfunction, polyendocrinopathy, enteropathy, X-linked syndrome), also known as X-linked autoimmunity-allergic dysregulation (XLAAD) syndrome. Transcript variants of FOXP3 encoding different isoforms have been identified. In human breast and colon cancer cells, expression of FOXP3 is regulated by p53 in response to the DNA damage.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: forkhead box P3; forkhead box P3 transcript variant; Forkhead box protein P3; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; JM2; MGC141961; MGC141963; regulatory protein Foxp3; RP23-54C14.1; Scurfin; unnamed protein product

          View more View less

          Gene Aliases: AIID; DIETER; FOXP3; IPEX; JM2; PIDX; XPID

          View more View less

          UniProt ID: (Human) Q9BZS1

          View more View less

          Entrez Gene ID: (Human) 50943, (Rhesus monkey) 574303

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding transcription corepressor activity protein binding zinc ion binding histone acetyltransferase binding identical protein binding protein homodimerization activity histone deacetylase binding sequence-specific DNA binding metal ion binding NF-kappaB binding NFAT protein binding sequence-specific double-stranded DNA binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter B cell homeostasis negative regulation of cytokine production myeloid cell homeostasis CD4-positive, CD25-positive, alpha-beta regulatory T cell differentiation CD4-positive, CD25-positive, alpha-beta regulatory T cell lineage commitment T cell mediated immunity tolerance induction tolerance induction to self antigen T cell tolerance induction regulation of immunoglobulin production positive regulation of T cell tolerance induction regulation of T cell anergy positive regulation of T cell anergy negative regulation of chronic inflammatory response negative regulation of T cell cytokine production positive regulation of peripheral T cell tolerance induction T cell anergy chromatin remodeling transcription, DNA-templated regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter inflammatory response negative regulation of cell proliferation response to virus gene expression positive regulation of gene expression negative regulation of gene expression establishment of endothelial blood-brain barrier negative regulation of NF-kappaB transcription factor activity response to lipopolysaccharide negative regulation of interferon-gamma production negative regulation of interleukin-10 production negative regulation of interleukin-17 production negative regulation of interleukin-2 production negative regulation of interleukin-4 production negative regulation of interleukin-5 production negative regulation of interleukin-6 production negative regulation of tumor necrosis factor production positive regulation of interleukin-4 production negative regulation of CREB transcription factor activity positive regulation of CD4-positive, CD25-positive, alpha-beta regulatory T cell differentiation transforming growth factor beta1 production positive regulation of transforming growth factor beta1 production immature T cell proliferation in thymus positive regulation of immature T cell proliferation in thymus CD4-positive, alpha-beta T cell proliferation T cell proliferation T cell activation negative regulation of T cell proliferation T cell homeostasis CD4-positive, alpha-beta T cell differentiation positive regulation of CD4-positive, alpha-beta T cell differentiation negative regulation of sequence-specific DNA binding transcription factor activity regulatory T cell differentiation positive regulation of regulatory T cell differentiation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter negative regulation of activated T cell proliferation alpha-beta T cell proliferation negative regulation of alpha-beta T cell proliferation lymphocyte proliferation isotype switching to IgE isotypes negative regulation of isotype switching to IgE isotypes regulation of isotype switching to IgG isotypes negative regulation of lymphocyte proliferation negative regulation of defense response to virus negative regulation of inflammatory response negative regulation of immune response T cell receptor signaling pathway response to rapamycin negative regulation of T-helper 17 cell differentiation negative regulation of CD4-positive, alpha-beta T cell proliferation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline