Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • T-bet Antibodies

          Invitrogen

          T-bet Monoclonal Antibody (eBio4B10 (4B10)), eBioscience™

          Advanced Verification
          47 Published Figures
          119 References
          View all (33) T-bet antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite T-bet Monoclonal Antibody (eBio4B10 (4B10)), eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          • Published Figures (47)
          • Advanced Verification (2)
          T-bet Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          T-bet Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 53

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          T-bet Antibody (14-5825-82) in ICC/IF

          Immunofluorescence analysis of T-bet was performed using 70% confluent log phase NK-92 cells. The cells were fixed with 4% paraformaldehyde for 10 Minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled with T-bet Monoclonal Antibody (Product # 14-5825-82) at 5 µg/mL in 0.1% BSA, incubated at 4 degree Celsius overnight and then labeled with Goat anti-Mouse IgG (H+L), Superclonal™ Recombinant Secondary Antibody, Alexa Fluor 488 (Product # A28175) at a dilu... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          T-bet Antibody in Immunocytochemistry (ICC/IF)
          T-bet Antibody in Western Blot (WB)
          T-bet Antibody in Western Blot (WB)
          T-bet Antibody in Western Blot (WB)
          T-bet Antibody in Immunocytochemistry (ICC/IF)
          T-bet Antibody in Immunohistochemistry (IHC)
          T-bet Antibody in Western Blot (WB)
          T-bet Antibody in Western Blot (WB)
          T-bet Antibody in Western Blot (WB)
          T-bet Antibody in Western Blot (WB)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody in Flow Cytometry (Flow)
          T-bet Antibody
          T-bet Antibody

          Product Details

          14-5825-82

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2 µg/mL
          View 8 publications 8 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-Dependent
          -

          Immunocytochemistry (ICC/IF)

          5 µg/mL
          View 5 publications 5 publications

          Flow Cytometry (Flow)

          -
          View 101 publications 101 publications

          Immunoprecipitation (IP)

          Assay-Dependent
          -

          ChIP assay (ChIP)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rhesus monkey

          Published species

          Bat, Human, Mouse, Rhesus monkey

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBio4B10 (4B10)

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 660 
          • APC 
          • eFluor 450
          • eFluor 660
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PerCP-Cyanine5.5
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_763634

          Product Specific Information

          Description: The eBio4B10 monoclonal antibody reacts with mouse and human T-bet. T-bet is a Th1-specific T-box transcription factor critical to the development of the Th1 CD4+ T cell lineage. This is known based on the observations that T-Bet deficient mice have impaired Th1 cell development, and that ectopic expression of T-Bet results in development skewed to the Th1 lineage. T-Bet expression is induced by the Th1 cytokine IFN gamma, and T-Bet also regulates the expression of IFN gamma, likely, at least in part, through the modification of DNA accessibility and histone remodeling. In addition to IFN gamma, T-Bet is also known to regulate the expression of IL-12R beta and IL-2. Moreover, T-Bet plays a role in class-switch recombination in B-cells.

          Applications Reported: This eBio4B10 antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded tissue sections (with citrate antigen retrieval), immunoprecipitation, and immunoblotting (WB).

          Applications Tested: This eBio4B10 antibody has been tested by western blot analysis of Th1-polarized mouse CD4+ T cells and can be used at 2 µg/mL as a starting dilution for western blotting.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          T-bet, also known as TBX21, is a T-cell specific transcription factor containing a single T-box DNA-binding domain. It controls the expression of the TH1 cytokine, interferon-gamma. Both Th1 and Th17 cells are crucial in immune regulation and autoimmune disease development. T-bet initiates Th1 lineage development from naive Th precursor cells and directs T cell differentiation to Th1 versus Th17. T-bet is cooperating with Stat4 (signal tranducer and activator of transcription 4) in programming chromatin architecture for Th1 gene expression. Genetic variations in T-bet are associated with susceptibility to aspirin-induced asthma and nasal polyps.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: HGNC:11599; T-box 21; T-box expressed in T cells; T-box protein 21; T-box transcription factor expressed in T cells; T-box transcription factor TBX21; T-cell-specific T-box transcription factor T-bet; Transcription factor TBLYM

          View more View less

          Gene Aliases: IMD88; T-bet; T-PET; TBET; TBLYM; TBT1; TBX21

          View more View less

          UniProt ID: (Human) Q9UL17, (Mouse) Q9JKD8

          View more View less

          Entrez Gene ID: (Human) 30009, (Rhesus monkey) 694044, (Mouse) 57765

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding sequence-specific DNA binding sequence-specific double-stranded DNA binding transcription regulatory region DNA binding Rel homology transcription factor
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter cell fate specification T-helper 1 cell lineage commitment regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter response to virus positive regulation of gene expression T cell differentiation negative regulation of interleukin-2 production proteasome-mediated ubiquitin-dependent protein catabolic process regulation of T cell differentiation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of isotype switching to IgG isotypes regulation of immune response lymphocyte migration negative regulation of T-helper 17 cell differentiation negative regulation of T-helper 17 cell lineage commitment negative regulation of T-helper 2 cell cytokine production positive regulation of T-helper 1 cell cytokine production transcription, DNA-templated cellular response to organic substance
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline