Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: PGCISLNRT SSQNISLPEY FSEHFHEPFP EGFSKPAEFT QHKFVKICME EPRFQANFPQ TWLWISGPLC LYCAERLYRY IRSNKPVTII SVISHPSDVM EIRMVKENFK ARPGQYITLH CPSVSALENH PFTLTMCPTE TKATFGVHLK IVGDWTERFR DLLLPPSSQD SEILPFIQSR NYPKLYIDGP FGSPFEESLN YEVSLCVAGG IGVTPFASIL NTLLDDWKPY KLRRLYFIWV CRDIQSFRWF ADLLCMLHNK FWQENRPDYV NIQLYLSQTD GIQKIIGEKY HALNSRLFIG RPRWKLLFDE IAKYNRGKTV GVFCCGPNSL SKTLHKLSNQ NNSYGTRFEY NKESFS (224-578 aa encoded by BC040105)
Oxygen sensing is essential for homeostasis in all aerobic organisms. A phagocyte-type oxidase, similar to that responsible for the production of large amounts of reactive oxygen species (ROS) in neutrophil granulocytes, with resultant antimicrobial activity, has been postulated to function in the kidney as an oxygen sensor that regulates the synthesis of erythropoietin in the renal cortex. NOX4 has a role as a redox messenger in the activation of intracellular signaling pathways leading (or contributing) to mitochondriogenesis, cell survival, and differentiation in hematopoietic stem cells. Data suggests that NOX4 provides a novel link between the insulin receptor and the generation of cellular reactive oxygen species that enhances insulin signal transduction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kidney oxidase-1; Kidney superoxide-producing NADPH oxidase; KOX-1; KOX1; NADPH oxidase 4; NOX 4; NOX-4; NOX4; NOX4v; Renal NAD(P)H-oxidase; Superoxide-generating NADPH oxidase 4; unnamed protein product
Gene Aliases: AI648021; KOX; KOX-1; NOX4; RENOX
UniProt ID: (Human) Q9NPH5, (Rat) Q924V1, (Mouse) Q9JHI8
Entrez Gene ID: (Human) 50507, (Rat) 85431, (Mouse) 50490
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support