Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • RAB11A/RAB11B Antibodies

          Proteintech

          RAB11A/B Polyclonal Antibody

          33 References
          View all (3) RAB11A/RAB11B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RAB11A/B Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (11)
          RAB11A/B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RAB11A/B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RAB11A/B Antibody (15903-1-AP) in ICC/IF

          Immunofluorescent analysis of (-20°C Methanol) fixed A549 cells using RAB11A/B antibody (Product # 15903-1-AP) at dilution of 1:400 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RAB11A/B Antibody in Immunocytochemistry (ICC/IF)
          RAB11A/B Antibody in Immunocytochemistry (ICC/IF)
          RAB11A/B Antibody in Immunocytochemistry (ICC/IF)
          RAB11A/B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB11A/B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB11A/B Antibody in Western Blot (WB)
          RAB11A/B Antibody in Western Blot (WB)
          RAB11A/B Antibody in Western Blot (WB)
          RAB11A/B Antibody in Western Blot (WB)
          RAB11A/B Antibody in Western Blot (WB)
          RAB11A/B Antibody in Immunoprecipitation (IP)
          RAB11A/B Polyclonal Antibody

          Product Details

          15903-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 13 publications 13 publications

          Immunohistochemistry (IHC)

          -
          View 5 publications 5 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 11 publications 11 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 2 publications 2 publications

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Dog, Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RAB11A/B Fusion Protein Ag8742 (1-216 aa encoded by BC013348)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.27 mg/mL

          Amount

          40.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody can recognize both RAB11A (24 kDa) and RAB11B(24 kDa).

          Immunogen sequence: MGTRDDEYD YLFKVVLIGD SGVGKSNLLS RFTRNEFNLE SKSTIGVEFA TRSIQVDGKT IKAQIWDTAG QERYRAITSA YYRGAVGALL VYDIAKHLTY ENVERWLKEL RDHADSNIVI MLVGNKSDLR HLRAVPTDEA RAFAEKNGLS FIETSALDST NVEAAFQTIL TEIYRIVSQK QMSDRRENDM SPSNNVVPIH VPPTTENKPK VQCCQNI (1-216 aa encoded by BC013348)

          Target Information

          Rab11A is a member of the YPT-1 subfamily of Ras-related GTP-binding proteins. Rab11A mRNA has been detected in a variety of tissues including brain, testis, spleen, heart, and gastrointestinal mucosa. In the gastric fundus the Rab11A protein is expressed in parietal cell tubulovesicles membranes. Expression of the Rab11A protein appears to occur in discrete epithelial cell populations where it localizes to apical vesicular populations. Recent studies have shown Rab11A is essential for resecretion of alpha-synuclein out of neuronal cells via endosome recycling and is required for exocytosis of discoidal/fusiform vesicles of bladder umbrella cells. Rab11A is also implicated in biogenesis of Birbeck granules and endosomal activation of the PI3K/AKT signaling pathway via G beta gamma trafficking.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: GTP-binding protein YPT3; GTPase; RAB 11A, member oncogene family; Rab-11; RAB11A; RAB11B; RAB11B, member of RAS oncogene family; RAS oncogene family protein; Ras-related protein Rab-11A; Ras-related protein Rab-11B; unnamed protein product; YL8; YPT3

          View more View less

          Gene Aliases: A730055L17Rik; H-YPT3; NDAGSCW; RAB11; RAB11A; RAB11B; YL8; YPT3

          View more View less

          UniProt ID: (Human) P62491, (Human) Q15907, (Mouse) P62492, (Mouse) P46638

          View more View less

          Entrez Gene ID: (Human) 8766, (Human) 9230, (Mouse) 53869, (Mouse) 19326

          View more View less

          Function(s)
          nucleotide binding GTPase activity obsolete small monomeric GTPase activity protein binding GTP binding microtubule binding hydrolase activity syntaxin binding myosin V binding dynein light intermediate chain binding GDP binding cadherin binding
          Process(es)
          cytokinesis transport intracellular protein transport exocytosis cell cycle mitotic metaphase plate congression small GTPase mediated signal transduction metabolic process positive regulation of epithelial cell migration regulation of multivesicular body size positive regulation of G2/M transition of mitotic cell cycle protein transport vesicle-mediated transport astral microtubule organization neuron projection development melanosome transport Rab protein signal transduction multivesicular body assembly positive regulation of axon extension regulation of long-term neuronal synaptic plasticity regulation of protein transport establishment of vesicle localization regulation of vesicle-mediated transport establishment of protein localization to organelle protein localization to plasma membrane establishment of protein localization to membrane mitotic spindle assembly exosomal secretion mitotic cytokinesis vesicle docking involved in exocytosis microtubule-based movement regulation of cytokinesis protein localization to cell surface Golgi to plasma membrane protein transport positive regulation of cilium assembly plasma membrane to endosome transport early endosome to recycling endosome transport protein localization to cilium protein transmembrane transport vesicle-mediated transport to the plasma membrane neurotransmitter receptor transport, endosome to postsynaptic membrane vesicle-mediated transport in synapse synaptic vesicle endosomal processing amyloid-beta clearance by transcytosis regulation of cilium assembly regulation of early endosome to recycling endosome transport positive regulation of protein localization to plasma membrane positive regulation of mitotic cytokinetic process regulation of protein localization to centrosome protein localization to cleavage furrow regulation of endocytic recycling receptor recycling endocytic recycling transferrin transport insulin secretion involved in cellular response to glucose stimulus regulation of anion transport constitutive secretory pathway regulated exocytosis cellular response to acidic pH regulation of protein localization to cell surface endocytosis retrograde transport, endosome to plasma membrane
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline