Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • CD90 (Thy1) Antibodies

          Invitrogen

          CD90 (Thy-1) Monoclonal Antibody (eBio5E10 (5E10)), APC-eFluor™ 780, eBioscience™

          48 Published Figures
          44 References
          View all (80) CD90 (Thy1) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD90 (Thy-1) Monoclonal Antibody (eBio5E10 (5E10)), APC-eFluor™ 780, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (48)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 49

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD90 (Thy-1) Antibody (47-0909-42) in Flow

          HEL cells were stained with Mouse IgG1 kappa Isotype Control, APC-eFluor 780 (Product # 47-4714-82) (blue histogram) or CD90 (Thy-1) Monoclonal Antibody, APC-eFluor 780 (purple histogram). Viable cells were used for analysis, as determined by Fixable Viability Dye eFluor 450 (Product # 65-0863-18). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
          CD90 (Thy-1) Antibody in Flow Cytometry (Flow)

          Product Details

          47-0909-42

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          5 µL (0.25 µg)/test
          View 44 publications 44 publications
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1, kappa

          Recommended Isotype Control

          Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), APC-eFluor™ 780, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBio5E10 (5E10)

          Conjugate

          APC-eFluor™ 780 APC-eFluor™ 780 APC-eFluor™ 780
          • Unconjugated
          • Alexa Fluor 700 
          • APC 
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 563
          • Brilliant Violet 480
          • Brilliant Violet 650
          • Brilliant Violet 786
          • FITC
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PerCP-Cyanine5.5
          • PerCP-eFluor 710
          • Super Bright 436
          • Request custom conjugation

          Excitation/Emission Max

          756/785 nm View spectra spectra

          Form

          Liquid

          Concentration

          5 µL/Test

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2, with BSA

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2848361

          Product Specific Information

          Description: The eBio5E10 monoclonal antibody reacts with human CD90, also known as Thy-1 (thymus cell antigen-1). CD90 is a 25-35 kD receptor expressed on thymocytes, CD34+ prothymocytes, hematopoietic stem cells, neurons, a small subset of human fetal liver cells, cord blood cells, and bone marrow cells. CD90 is expressed on a subset of immature, CD34+ cells and a distinct subset of mature CD34- cells that are CD3+CD4+. The CD90+CD34+ population is enriched for cells capable of long-term culture. CD90 is involved in regulation of adhesion and signal transduction by T cells.

          Applications Reported: This eBio5E10 (5E10) antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This eBio5E10 (5E10) antibody has been pre-diluted and tested by flow cytometric analysis of human erythroleukemia (HEL) cells. This may be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

          APC-eFluor 780 emits at 780 nm and is excited with the Red laser (633-647 nm). Please make sure that your instrument is capable of detecting this fluorochrome.

          Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.

          Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222-49) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-57) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

          Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser.

          Target Information

          CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. CD90 is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. CD90 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain. CD90 can be used as a marker for a variety of stem cells and for the axonal processes of mature neurons. Structural study of CD90 led to the foundation of the Immunoglobulin superfamily, of which it is the smallest member, and led to the first biochemical description and characterization of a vertebrate GPI anchor. Diseases associated with CD90 dysfunction include nasopharyngeal carcinoma and thymoma.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: CD90; CDw90; FLJ33325; Thy-1 antigen; Thy-1 membrane glycoprotein; Thy-1 T-cell antigen; unnamed protein product

          View more View less

          Gene Aliases: CD90; CDw90; THY1

          View more View less

          UniProt ID: (Human) P04216

          View more View less

          Entrez Gene ID: (Human) 7070

          View more View less

          Function(s)
          GTPase activator activity integrin binding protein binding GPI anchor binding
          Process(es)
          angiogenesis regulation of cell-matrix adhesion regulation of immune system process positive regulation of cellular extravasation negative regulation of protein kinase activity cytoskeleton organization cell adhesion cell surface receptor signaling pathway integrin-mediated signaling pathway cell-cell signaling regulation of cell migration negative regulation of cell migration heterotypic cell-cell adhesion receptor clustering positive regulation of GTPase activity positive regulation of cell adhesion retinal cone cell development focal adhesion assembly negative regulation of axonogenesis T cell receptor signaling pathway negative regulation of T cell receptor signaling pathway positive regulation of T cell activation positive regulation of release of sequestered calcium ion into cytosol positive regulation of focal adhesion assembly negative regulation of neuron projection regeneration cell-cell adhesion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline