Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: LLNLYSLVYN EDKKNTMGPA LERIDYLLIL WIIGMIWSDI KRLWYEGLED FLEESRNQLS FVMNSLYLAT FALKVVAHNK FHDFADRKDW DAFHPTLVAE G
Thought to form a receptor-activated non-selective calcium permeant cation channel. Probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. Seems to be also activated by intracellular calcium store depletion. Interacts with ITPR3. Appears to be ubiquitous.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: capacitative calcium channel protein Trp1; Short transient receptor potential channel 1; transient receptor potential canonical 1; transient receptor potential; Ca2+ permeable ion channel; Transient receptor protein 1; TRP-1; Trp-related protein 1; TRPC1
Gene Aliases: HTRP-1; Mtrp1; TRP1; TRPC1; Trrp1
UniProt ID: (Human) P48995, (Mouse) Q61056, (Rat) Q9QX01
Entrez Gene ID: (Human) 7220, (Mouse) 22063, (Rat) 89821
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support