Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • SHANK3 Antibodies

          Invitrogen

          SHANK3 Monoclonal Antibody (S69), PerCP

          View all (28) SHANK3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SHANK3 Monoclonal Antibody (S69), PerCP

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          SHANK3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SHANK3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SHANK3 Antibody (MA5-45460) in ICC/IF

          Immunocytochemistry/Immunofluorescence analysis using human neuroblastoma cells. Fixation involved 4% PFA for 15 min. Samples were incubated with SHANK3 monoclonal antibody (Product # MA5-45460) at 1:50 for overnight at 4°C with slow rocking, followed by AlexaFluor 488 at 1:1,000 for 1 hour at RT. Counterstain used was Phalloidin-iFluor 647 (red) F-Actin stain; Hoechst (blue) nuclear stain at 1:800, 1.6mM for 20 min at RT. (A) Hoechst (blue) nuclear stain. (B) Phalloidin-iFluor 647 (red) F-Actin stain. (C) SHANK3 antibody (D) Composite. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SHANK3 Antibody in Immunocytochemistry (ICC/IF)
          SHANK3 Antibody in Immunocytochemistry (ICC/IF)
          SHANK3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SHANK3 Antibody in Western Blot (WB)
          SHANK3 Monoclonal Antibody (S69), PerCP

          Product Details

          MA5-45460

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100
          -

          Immunocytochemistry (ICC/IF)

          1:100
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -

          Antibody Microarray (Ab Microarray)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          S69

          Immunogen

          Synthetic peptide amino acids 840-857 of rat Shank3
          3D Epitope / Immunogen

          Conjugate

          PerCP PerCP PerCP
          • Unconjugated
          • APC 
          • FITC
          • PE 
          • Request custom conjugation

          Excitation/Emission Max

          482/675 nm View spectra spectra

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          95.64mM phosphate/2.48mM MES, pH 7.4, with 0.5M EDTA

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2931914

          Product Specific Information

          1 µg/mL of MA5-45460 was sufficient for detection of Shank3 in 10 µg COS cell lysate transiently transfected with Shank3 by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 190kDa. No cross-reactivity against Shank1 or Shank2.

          Target Information

          SHANK3 is a major scaffold postsynaptic density protein which interacts with multiple proteins and complexes to orchestrate the dendritic spine and synapse formation, maturation and maintenance. It interconnects receptors of the postsynaptic membrane including NMDA-type and metabotropic glutamate receptors via complexes with GKAP/PSD-95 and HOMER, respectively, and the actin-based cytoskeleton. SHANK3 plays a role in the structural and functional organization of the dendritic spine and synaptic junction through the interaction with Arp2/3 and WAVE1 complex as well as the promotion of the F-actin clusters. By way of this control of actin dynamics, SHANK3 participates in the regulation of developing neurons growth cone motility and the NMDA receptor-signaling. It also modulates GRIA1 exocytosis and GRM5/MGLUR5 expression and signaling to control the AMPA and metabotropic glutamate receptor-mediated synaptic transmission and plasticity. SHANK3 may be required at an early stage of synapse formation and be inhibited by IGF1 to promote synapse maturation. (Uniprot)

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: proline rich synapse associated protein 2; Proline-rich synapse-associated protein 2; ProSAP2; SH3 and multiple ankyrin repeat domains protein 3; SH3 and multiple ankyrin repeat domains protein 3 variant c-3; SH3 and multiple ankyrin repeat domains protein 3 variant c-4; SH3 and multiple ankyrin repeat domains-like protein 3 variant 1; SH3 and multiple ankyrin repeat domains-like protein 3 variant 2; SH3 and multiple ankyrin repeat domains-like protein 3 variant 3; SH3 and multiple ankyrin repeat domains-like protein 3 variant c-1; SH3 and multiple ankyrin repeat domains-like protein 3 variant c-2; SH3 and multiple ankyrin repeat domains-like protein 3 variant d-1; SH3 and multiple ankyrin repeat domains-like protein 3 variant d-2; SH3 and multiple ankyrin repeat domains-like protein 3 variant d-3; SH3 and multiple ankyrin repeat domains-like protein 3 variant e-1; SH3 and multiple ankyrin repeat domains-like protein 3 variant e-2; SH3/ankyrin domain gene 3; shank postsynaptic density protein; Shank postsynaptic density protein 3a; Shank3; shank3 postsynaptic density protein; shank3 splice variant II; shank3 splice variant III; shank3 splice variant IV; shank3e splice variant III; shank3e splice variant IV; SPANK 2; SPANK-2; SPANK2

          View more View less

          Gene Aliases: AI841104; DEL22q13.3; KIAA1650; PROSAP2; PSAP2; SCZD15; SHANK3; Shank3b; SHANK3c-3; SHANK3c-4; SPANK-2

          View more View less

          UniProt ID: (Human) Q9BYB0, (Mouse) Q4ACU6, (Rat) Q9JLU4

          View more View less

          Entrez Gene ID: (Human) 85358, (Mouse) 58234, (Rat) 59312

          View more View less

          Function(s)
          actin binding protein binding zinc ion binding SH3 domain binding GKAP/Homer scaffold activity ionotropic glutamate receptor binding scaffold protein binding protein C-terminus binding identical protein binding protein self-association
          Process(es)
          MAPK cascade embryonic epithelial tube formation synapse assembly learning memory striatal medium spiny neuron differentiation adult behavior negative regulation of actin filament bundle assembly social behavior locomotory exploration behavior vocal learning negative regulation of cell volume positive regulation of long-term neuronal synaptic plasticity brain morphogenesis neuromuscular process controlling balance protein oligomerization positive regulation of synapse structural plasticity positive regulation of synaptic transmission, glutamatergic dendritic spine morphogenesis positive regulation of dendritic spine development regulation of dendritic spine morphogenesis vocalization behavior postsynaptic density assembly AMPA glutamate receptor clustering NMDA glutamate receptor clustering guanylate kinase-associated protein clustering regulation of long-term synaptic potentiation positive regulation of long-term synaptic potentiation positive regulation of glutamate receptor signaling pathway regulation of long term synaptic depression positive regulation of excitatory postsynaptic potential regulation of grooming behavior regulation of behavioral fear response positive regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity associative learning synapse organization cognition
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline