Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Antibody detects endogenous levels of total MAGE1.
This gene encodes an alpha-dystrobrevin-associated MAGE (melanoma-associated antigen) protein, which is a member of the MAGE family. The protein contains a nuclear localization signal in the N-terminus, 30 12-amino acid repeats beginning at nt 60 with the consensus sequence ASEGPSTSVLPT, and two MAGE domains in the C-terminus. It may play a signaling role in brain, muscle, and peripheral nerve. This gene is located on X chromosome in a region containing loci linked to cognitive disability.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-dystrobrevin-associated MAGE Protein; DAMAGE; dystrobrevin-associated MAGE protein; hepatocellular carcinoma-associated HCA1; Hepatocellular carcinoma-associated protein 1; hepatocellular carcinoma-associated protein HCA1; MAGE-E1 antigen; melanoma antigen family E, 1; melanoma antigen family E1; Melanoma-associated antigen E1; unnamed protein product
Gene Aliases: AI847422; DAMAGE; HCA1; KIAA1587; MAGEE1; mMage-e1; RGD1560259
UniProt ID: (Human) Q9HCI5, (Mouse) Q6PCZ4, (Rat) A1A5P9
Entrez Gene ID: (Human) 57692, (Mouse) 107528, (Rat) 317232
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support