Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 5
Positive Control: Rat brain tissue lysate, human brain tissue lysate, mouse brain tissue lysates, rat brain tissue, mouse brain tissue, SH-SY5Y.
Subcellular Location: Nucleus membrane, Cytoplasm.
The protein encoded by this gene catalyzes the formation of inositol 1,4,5-trisphosphate and diacylglycerol from phosphatidylinositol 4,5-bisphosphate. This reaction uses calcium as a cofactor and plays an important role in the intracellular transduction of many extracellular signals. This gene is activated by two G-protein alpha subunits, alpha-q and alpha-11.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-phosphatidyl-D-myo-inositol-4,5-bisphosphate; 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-1; inositoltrisphosphohydrolase; monophosphatidylinositol phosphodiesterase; phosphoinositidase C; phosphoinositide phospholipase C; Phosphoinositide phospholipase C-beta-1; phosphoinositide-specific phospholipase c; phospholipase C, beta 1 (phosphoinositide-specific); Phospholipase C-beta-1; phospholipase C-beta-1a; Phospholipase C-beta1; Phospholipase C-I; PLC-154; PLC-beta-1; PLC-I; triphosphoinositide phosphodiesterase; unnamed protein product
Gene Aliases: 3110043I21Rik; AI132408; DEE12; EIEE12; KIAA0581; mKIAA0581; Phosphb; PI-PLC; PLC'1; PLC-154; PLC-beta-1; PLC-I; PLC154; Plcb; PLCB1; PLCB1A; PLCB1B; PLCbeta1; RATPHOSPHB
UniProt ID: (Human) Q9NQ66, (Rat) P10687, (Mouse) Q9Z1B3
Entrez Gene ID: (Human) 23236, (Rat) 24654, (Mouse) 18795
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support