Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody is predicted to react with rabbit based on sequence homology.
FUT1 is a Golgi stack membrane protein that is involved in the creation of a precursor of the H antigen, which is required for the final step in the soluble A and B antigen synthesis pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a-1, 2 fucosyltransferase (H antigen); alpha(1,2) fucosyltransferase 1; Alpha(1,2)FT 1; Blood group H alpha 2-fucosyltransferase; Fucosyltransferase 1; galactoside 2-alpha-L-fucosyltransferase; galactoside 2-L-fucosyltransferase enzyme; Galactoside alpha-(1,2)-fucosyltransferase 1; GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase; GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 1; H antigen; H glycosyltransferase; H glycosytransferase; nonfunctional fucosyltransferase 1; Type 1 galactoside alpha-(1,2)-fucosyltransferase FUT1; Type 2 galactoside alpha-(1,2)-fucosyltransferase FUT1
Gene Aliases: FUT1; H; HH; HSC
UniProt ID: (Human) P19526
Entrez Gene ID: (Human) 2523
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support