Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 8
Recommended positive controls: 293T, A431, HeLa, HepG2, mouse brain.
Predicted reactivity: Mouse (96%), Rat (95%), Xenopus laevis (90%), Pig (96%), Chicken (82%), Rhesus Monkey (99%), Bovine (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ATP6V1A encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c", and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is one of two V1 domain A subunit isoforms and is found in all tissues. Transcript variants derived from alternative polyadenylation exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 70-kDa subunit; ATPase, H+ transporting, lysosomal (vacuolar proton pump), alpha 70 kDa, isoform 2; ATPase, H+ transporting, lysosomal 70kD, V1 subunit A, isoform 1; ATPase, H+ transporting, lysosomal 70kDa, V1 subunit A; ATPase, H+ transporting, lysosomal, subunit A1; ATPase, H+ transporting, V1 subunit A, isoform 1; ATPase, H+ transporting, V1 subunit A1; H(+)-transporting two-sector ATPase, subunit A; H+-transporting ATPase chain A, vacuolar (VA68 type); lysosomal 70kDa; unnamed protein product; V-ATPase 69 kDa subunit; V-ATPase 69 kDa subunit 1; V-ATPase A subunit 1; V-ATPase subunit A; V-type proton ATPase (V-ATPase) catalytic subunit A; V-type proton ATPase catalytic subunit A; Vacuolar ATPase isoform VA68; vacuolar proton pump alpha subunit 1; Vacuolar proton pump subunit alpha; vacuolar-type H(+)-ATPase
Gene Aliases: AI647066; ARCL2D; ATP6A1; Atp6a2; ATP6V1A; ATP6V1A1; DEE93; HO68; IECEE3; VA68; Vma1; VPP2
UniProt ID: (Human) P38606, (Mouse) P50516
Entrez Gene ID: (Human) 523, (Mouse) 11964, (Rat) 685232
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support