Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MFVSLWEFFY GHFFRFWMKW LLRQMTGKCE LQRIFDTYGG AQRTYRIENS LTYSKNKVLQ NATRVAQSEL DRCIANIMKE KNICSEKDTS FQICMRTCLL QITGYKQLYH DVENVRKKPY DSANAQHEKM
Positive Samples: U-87MG, LO2
This gene encodes one of six engulfment and motility (ELMO) domain-containing proteins. This gene is thought to play a role in antiviral responses. Mutations in this gene may be involved in the cause of familial idiopathic pulmonary fibrosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ELMO domain containing 2; ELMO domain-containing protein 2; ELMO/CED-12 domain containing 2; unnamed protein product
Gene Aliases: 9830169G11Rik; ELMOD2
UniProt ID: (Human) Q8IZ81, (Mouse) Q8BGF6
Entrez Gene ID: (Human) 255520, (Mouse) 244548
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support