Search
Search

Sipa1| Gene Symbol: Sipa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_001164480.1 | NP_001157952.1 | ||||
| NM_001164481.1 | NP_001157953.1 | |||||
| NM_001164482.1 | NP_001157954.1 | |||||
| NM_001164568.1 | NP_001158040.1 | |||||
| NM_011379.4 | NP_035509.4 | |||||
| GenBank mRNA | AK028151.1 | - | ||||
| AK137578.1 | - | |||||
| AK149669.1 | - | |||||
| AK156280.1 | - | |||||
| AK169945.1 | - | |||||
| AK170246.1 | - | |||||
| AK178405.1 | - | |||||
| AK179748.1 | - | |||||
| AK196305.1 | - | |||||
| AK198804.1 | - | |||||
| AK198993.1 | - | |||||
| AK200374.1 | - | |||||
| AK205672.1 | - | |||||
| AK220472.1 | - | |||||
| BB843238.1 | - | |||||
| BC043679.1 | - | |||||
| BC054824.1 | - | 5324326 | ||||
| D11374.1 | - | |||||