Search
Search

ZMYM4| Gene Symbol: ZMYM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | XM_021095899.1 | XP_020951558.1 | ||||
| XM_021095900.1 | XP_020951559.1 | |||||
| XM_021095901.1 | XP_020951560.1 | |||||
| XM_021095902.1 | XP_020951561.1 | |||||
| XM_021095903.1 | XP_020951562.1 | |||||
| XM_021095904.1 | XP_020951563.1 | |||||
| XM_021095905.1 | XP_020951564.1 | |||||
| XM_021095906.1 | XP_020951565.1 | |||||
| XM_021095907.1 | XP_020951566.1 | |||||
| XM_021095908.1 | XP_020951567.1 | |||||
| XM_021095909.1 | XP_020951568.1 | |||||
| XM_021095910.1 | XP_020951569.1 | |||||
| GenBank mRNA | AK233972.1 | - | ||||
| AK348051.1 | - | |||||