Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFE2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624612.3 | XP_005624669.1 | 1615 | 82 | ||
XM_005624613.3 | XP_005624670.1 | 1525 | 82 | |||
XM_014116620.2 | XP_013972095.1 | 1502 | 82 | |||
XM_022423058.1 | XP_022278766.1 | 1501 | 82 | |||
XM_846772.5 | XP_851865.1 | 1618 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris |