Search
Search

Pitx2| Gene Symbol: Pitx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514240.2 | XP_003514288.1 | 2 - 3 | 416 | 88 | |
| XM_003514242.2 | XP_003514290.1 | 3 - 4 | 356 | 88 | ||
| XM_007654423.1 | XP_007652613.1 | 3 - 4 | 395 | 88 | ||
Set Membership: |
|