Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ect | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274748.1 | NP_001261677.1 | 5 - 6 | 1011 | 86 | |
NM_001300090.1 | NP_001287019.1 | 6 - 7 | 1134 | 86 | ||
NM_079967.6 | NP_524706.2 | 6 - 7 | 1167 | 86 | ||
NM_168400.3 | NP_729596.1 | 6 - 7 | 1110 | 86 | ||
GenBank mRNA | AY061556.1 | - | 6 - 7 | 1151 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |