Search
Search

foxp3a| Gene Symbol: foxp3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001329567.1 | NP_001316496.1 | 6 - 7 | 600 | 79 | |
| XM_021478298.1 | XP_021333973.1 | 6 - 7 | 656 | 79 | ||
| GenBank mRNA | BC154484.1 | - | 6 - 7 | 600 | 79 | |
| BC163356.1 | - | 5 - 6 | 579 | 79 | ||
| FJ906821.1 | - | 5 - 6 | 506 | 79 | ||
| FM881778.1 | - | 5 - 6 | 573 | 79 | ||
Set Membership: |
|