Search
Search

adh8a| Gene Symbol: adh8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001946.4 | NP_001001946.3 | 9 | 1328 | 126 | |
| GenBank mRNA | AF295407.2 | - | 9 | 1257 | 126 | |
| BC065464.1 | - | 9 | 1251 | 126 | ||
| BC096855.1 | - | 9 | 1277 | 126 | ||
Set Membership: |
|