Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460855 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460863 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895838 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895875 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895896 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460829 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460847 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460903 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895831 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460812 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460897 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895929 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460830 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16573616 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895925 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460884 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895860 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895897 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895841 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895895 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460891 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895840 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895846 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895912 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895879 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895908 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895884 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460842 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895850 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460896 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460880 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895853 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460858 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895885 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895847 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895874 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895844 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460895 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460885 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895881 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460870 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460886 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460882 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895862 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460894 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895878 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460818 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895900 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460834 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895901 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895877 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460833 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895928 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895873 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895893 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460835 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895890 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895882 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895907 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895921 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895894 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895855 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895830 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895849 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895833 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460817 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460879 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895832 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895930 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16573624 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895889 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460889 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460877 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895861 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895856 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895923 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460892 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895888 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460890 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895845 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460827 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460840 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895870 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895903 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895899 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895837 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08460900 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28895835 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
30588 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
106759 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
8317 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
18275 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
531214 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
304359 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_24_P181149 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P95612 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_24_P181149 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
41670 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
37321 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P139853 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P128468 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P139853 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P128468 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
28742 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P33261032 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_24_P181149 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_24_P181149 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460879 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460877 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460900 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895932 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460890 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895899 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895901 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895903 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895912 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460885 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460897 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460889 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895929 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460895 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460891 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895921 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895908 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895923 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460894 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895928 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895925 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895907 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460890 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895923 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895928 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895907 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460897 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895903 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895901 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895932 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460889 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895925 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460879 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895929 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895908 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460885 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895899 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460891 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460900 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460877 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460894 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08460895 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895912 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28895921 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_24_P181149 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_23_P95612 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P139853 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P128468 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08460898 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08460902 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08460904 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08460878 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08460881 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08460876 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08460889 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08460887 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08460885 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08460872 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08460869 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08460891 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08460874 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08460899 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08460895 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08460866 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08460883 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16573620 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P08460893 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P08460889 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16573621 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P08460890 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P08460891 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P08460892 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P128468 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P139853 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P128468 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P139853 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P95612 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_24_P181149 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
159432 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
151904 |
AB051433.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
151904 |
AB079066.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
151904 |
AK025083.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
151904 |
AK026892.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
151904 |
AL137600.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
151904 |
AL832794.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
151904 |
BC004278.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
151904 |
BC008382.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
151904 |
BC108712.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
151904 |
NM_022766.5 |
refseq_mrna_human_NM |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
68357 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
91995 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P128468 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P139853 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P33261032 |
- |
- |
Agilent Human 2 cDNA Microarray (G4101A) |
4337387 |
- |
- |
Agilent Human 2 cDNA Microarray (G4101A) |
4337387 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460889 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460891 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460816 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460810 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460811 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460895 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460855 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460878 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460857 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460843 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460839 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460834 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460876 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460883 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460869 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460864 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460887 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16573613 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460831 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460890 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460849 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460861 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460862 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460818 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460836 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460844 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16573612 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460827 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460872 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460898 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460814 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460881 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460852 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460904 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460856 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16573616 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460821 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460900 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16573625 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460846 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460859 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460841 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460866 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460899 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460829 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460885 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460819 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460874 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460809 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460837 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08460825 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P139853 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P128468 |
- |
- |