Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145122.2 | NP_660104.2 | 5 - 6 | 789 | 62 | |
NR_028114.1 | 5 - 6 | 498 | 62 | |||
GenBank mRNA | AK020743.1 | - | 5 - 6 | 499 | 62 | |
AK143073.1 | - | 5 - 6 | 791 | 62 | ||
AK162716.1 | - | 5 - 6 | 489 | 62 | ||
BC010822.1 | - | 5 - 6 | 530 | 4218547 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |