Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161365.1 | NP_001154837.1 | 8 - 9 | 2555 | 83 | |
NM_177620.4 | NP_808288.2 | 8 - 9 | 2666 | 83 | ||
GenBank mRNA | AK032205.1 | - | 8 - 9 | 2667 | 83 | |
AK154506.1 | - | 8 - 9 | 2691 | 83 | ||
AK154522.1 | - | 8 - 9 | 2470 | 83 | ||
AK154963.1 | - | 8 - 9 | 2556 | 83 | ||
AK166196.1 | - | 8 - 9 | 2512 | 83 | ||
AK171301.1 | - | 8 - 9 | 2480 | 83 | ||
AK171311.1 | - | 8 - 9 | 2471 | 83 | ||
AY265356.1 | - | 8 - 9 | 2652 | 83 | ||
BC058432.1 | - | 2 - 3 | 309 | 83 | ||
BC137907.1 | - | 8 - 9 | 2594 | 8860926 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |